Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003790.3 | NP_001003790.1 | 6 | 667 | ||
NM_001003791.2 | NP_001003791.1 | 6 | 438 | |||
NM_007175.6 | NP_009106.1 | 6 | 438 | |||
XM_005273392.2 | XP_005273449.1 | 6 | 598 | |||
XM_006716280.2 | XP_006716343.1 | 5 | 343 | |||
XM_017013000.1 | XP_016868489.1 | 6 | 436 | |||
GenBank mRNA | AK291394.1 | - | 6 | 391 | ||
AK297279.1 | - | 6 | 391 | |||
AL442077.1 | - | 425 | ||||
AY358108.1 | - | 6 | 382 | |||
AY358851.1 | - | 6 | 372 | |||
BC005950.1 | - | 6 | 361 | |||
BC048308.1 | - | 6 | 661 | |||
BC050611.1 | - | 6 | 392 | |||
BC067765.1 | - | 6 | 665 | 5296776 | ||
BI560439.1 | - | 6 | 673 | |||
BP353279.1 | - | 6 | 438 |