Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENSA SIRNA Assays ›
          Gene Symbol:
          ENSA
          Gene Name:
          endosulfine alpha
          Gene Aliases:
          ARPP-19e
          Chromosome Location:
          Chr.1: 150622123 - 150629622 on Build GRCh38
          UniGene ID:
          Hs.632456
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s225853

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ENSA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004436.2 NP_004427.1 2 288
          NM_207042.1 NP_996925.1 2 288
          NM_207043.1 NP_996926.1 2 288
          NM_207044.1 NP_996927.1 2 288
          NM_207045.1 NP_996928.1 2 152
          NM_207046.1 NP_996929.1 2 152
          NM_207047.1 NP_996930.1 2 152
          NM_207168.1 NP_997051.1 2 288
          GenBank mRNA AF067170.1 - 2 245
          AF157509.1 - 2 242
          AF157510.1 - 2 242
          AK001981.1 - 2 262
          AK057673.1 - 1 2084
          AK290064.1 - 2 135
          AL711949.1 - 2 291
          BC000436.2 - 2 236 2820528
          BC004461.1 - 2 236 2820528
          BC068544.1 - 2 253 5273162
          BC069208.1 - 2 226 4853917
          BE799496.1 - 2 172
          BQ883543.1 - 2 250
          BU075511.1 - 2 213
          BU858280.1 - 146
          CR457130.1 - 2 136
          CR536578.1 - 2 136
          CR749580.1 - 2 243
          T78564.1 - 2 167
          X99906.1 - 2 261

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_021842, NM_001033974, FQ212889, FQ216114, FQ214207, and others.
          This siRNA also targets the following transcripts in Mouse: NM_019561, NM_001026212, BC059724, AK149726, AK006149, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          transport
          mitotic nuclear division
          response to nutrient
          response to glucose
          regulation of protein phosphatase type 2A activity
          negative regulation of catalytic activity
          regulation of insulin secretion
          cell division
          protein phosphatase inhibitor activity
          receptor binding
          protein binding
          ion channel inhibitor activity
          protein phosphatase type 2A regulator activity
          phosphatase inhibitor activity
          potassium channel inhibitor activity
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pqldv:80/100.66.72.197:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0