Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCC1 SIRNA Assays ›
Gene Symbol:
TMCC1
Gene Name:
transmembrane and coiled-coil domain family 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 129647792 - 129893605 on Build GRCh38
UniGene ID:
Hs.477547
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225924
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001017395.3
NP_001017395.2
5
1991
NM_001128224.2
NP_001121696.1
5
1392
NR_033361.1
3
1537
XM_006713542.3
XP_006713605.1
6
2104
XM_006713543.3
XP_006713606.1
6
2101
XM_006713544.1
XP_006713607.1
6
2229
XM_006713550.3
XP_006713613.1
6
1662
XM_006713552.3
XP_006713615.1
3
1284
XM_006713553.3
XP_006713616.1
3
2576
XM_011512572.2
XP_011510874.1
7
2264
XM_011512573.2
XP_011510875.1
6
2843
XM_011512576.2
XP_011510878.1
6
2097
XM_011512579.2
XP_011510881.1
5
1852
XM_011512580.2
XP_011510882.1
8
1989
XM_011512581.2
XP_011510883.1
7
1826
XM_011512582.1
XP_011510884.1
3
1618
XM_011512583.2
XP_011510885.1
5
1395
XM_011512584.2
XP_011510886.1
7
1714
XM_017005931.1
XP_016861420.1
6
2012
XM_017005932.1
XP_016861421.1
7
2251
XM_017005933.1
XP_016861422.1
5
2048
XM_017005934.1
XP_016861423.1
9
2062
XM_017005935.1
XP_016861424.1
10
2151
XM_017005936.1
XP_016861425.1
7
1825
XM_017005937.1
XP_016861426.1
7
1759
XM_017005938.1
XP_016861427.1
9
2125
XM_017005939.1
XP_016861428.1
8
2019
XM_017005940.1
XP_016861429.1
5
1397
XM_017005941.1
XP_016861430.1
5
1523
GenBank mRNA
AB018322.1
-
2
558
AK124671.1
-
3
1538
AK291448.1
-
5
1810
AK293408.1
-
3
1074
BC012480.1
-
2
121
BC030619.2
-
3
1455
BC034284.1
-
2
141
BC039859.1
-
5
1898
5527623
BC047517.1
-
3
1455
BX640712.1
-
3
1045
CR749206.1
-
3
1517
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0