Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KDM4C SIRNA Assays ›
Gene Symbol:
KDM4C
Gene Name:
lysine demethylase 4C
Gene Aliases:
GASC1, JHDM3C, JMJD2C, TDRD14C
Chromosome Location:
Chr.9: 6720863 - 7175648 on Build GRCh38
UniGene ID:
Hs.709425
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225929
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KDM4C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146695.1
NP_001140167.1
14
2581
NM_001146696.1
NP_001140168.1
14
2153
NM_001304339.1
NP_001291268.1
14
2581
NM_001304340.1
NP_001291269.1
12
2183
NM_015061.3
NP_055876.2
14
2581
NR_130707.1
15
3524
XM_006716741.2
XP_006716804.1
14
2250
XM_011517811.2
XP_011516113.1
15
2205
XM_011517812.2
XP_011516114.2
10
1395
XM_011517816.2
XP_011516118.1
7
1206
XM_017014498.1
XP_016869987.1
14
2250
XM_017014499.1
XP_016869988.1
16
2292
XM_017014500.1
XP_016869989.1
16
2319
XM_017014501.1
XP_016869990.1
13
1934
XM_017014502.1
XP_016869991.1
14
2250
XM_017014503.1
XP_016869992.1
14
2250
XM_017014504.1
XP_016869993.1
14
2250
XM_017014505.1
XP_016869994.1
6
936
XM_017014506.1
XP_016869995.1
15
2265
GenBank mRNA
AB018323.1
-
14
2161
AB037901.1
-
14
2151
AK296401.1
-
7
1251
AK303651.1
-
13
1991
AK304032.1
-
14
2079
AK304095.1
-
12
1756
AK310439.1
-
14
2164
AK314355.1
-
14
2146
BC104859.1
-
14
2061
8143862
BC104861.1
-
14
2061
8143864
BC143571.1
-
14
2061
9052082
BX645680.1
-
2
66
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
positive regulation of cell proliferation
histone H3-K9 demethylation
oxidation-reduction process
zinc ion binding
enzyme binding
histone demethylase activity
histone demethylase activity (H3-K9 specific)
androgen receptor binding
dioxygenase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0