Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281748.1 | NP_001268677.1 | 22, 23 | 3612 | ||
NM_001281749.1 | NP_001268678.1 | 21, 22 | 3488 | |||
NM_025164.4 | NP_079440.3 | 21, 22 | 3668 | |||
XM_005271481.2 | XP_005271538.1 | 22, 23 | 3850 | |||
XM_005271482.4 | XP_005271539.2 | 21, 22 | 3706 | |||
XM_005271484.3 | XP_005271541.2 | 21, 22 | 3526 | |||
XM_005271485.3 | XP_005271542.1 | 9, 10 | 2188 | |||
XM_011542721.2 | XP_011541023.1 | 22, 23 | 3812 | |||
XM_011542722.1 | XP_011541024.1 | 22, 23 | 3670 | |||
XM_011542723.2 | XP_011541025.1 | 22, 23 | 3693 | |||
XM_011542724.2 | XP_011541026.1 | 21, 22 | 3547 | |||
XM_011542725.2 | XP_011541027.1 | 20, 21 | 3430 | |||
XM_017017424.1 | XP_016872913.1 | 20, 21 | 3382 | |||
XM_017017425.1 | XP_016872914.1 | 22, 23 | 3888 | |||
XM_017017426.1 | XP_016872915.1 | 20, 21 | 3762 | |||
XM_017017427.1 | XP_016872916.1 | 17, 18 | 3165 | |||
GenBank mRNA | AB209327.1 | - | 14, 15 | 2678 | ||
AK022302.1 | - | 3, 4 | 1223 | |||
AK122838.1 | - | 7, 8 | 2930 | |||
AY598338.1 | - | 3, 4 | 1220 | |||
BC063887.1 | - | 7, 8 | 1681 | |||
BC086858.1 | - | 3, 4 | 1223 | |||
BC128510.1 | - | 21, 22 | 3350 | 40081639 | ||
BC128511.1 | - | 22, 23 | 3468 | 40081640 |