Search
Search

CFAP65| Gene Symbol: CFAP65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278295.1 | NP_001265224.1 | 5 | 478 | ||
| NM_001278296.1 | NP_001265225.1 | 4 | 429 | |||
| NM_152389.3 | NP_689602.2 | 4 | 429 | |||
| NM_194302.3 | NP_919278.2 | 5 | 584 | |||
| XM_011510903.1 | XP_011509205.1 | 5 | 645 | |||
| XM_011510904.2 | XP_011509206.1 | 5 | 2229 | |||
| XM_011510908.1 | XP_011509210.1 | 5 | 551 | |||
| XM_011510909.1 | XP_011509211.1 | 5 | 591 | |||
| XM_011510910.1 | XP_011509212.1 | 5 | 645 | |||
| XM_011510911.1 | XP_011509213.1 | 5 | 496 | |||
| XM_011510912.1 | XP_011509214.1 | 5 | 539 | |||
| XM_011510914.1 | XP_011509216.1 | 4 | 436 | |||
| XM_011510915.1 | XP_011509217.1 | 4 | 398 | |||
| XM_011510916.1 | XP_011509218.1 | 5 | 645 | |||
| XM_011510919.1 | XP_011509221.1 | 5 | 645 | |||
| XM_017003752.1 | XP_016859241.1 | 6 | 712 | |||
| XM_017003753.1 | XP_016859242.1 | 5 | 2069 | |||
| XM_017003754.1 | XP_016859243.1 | 4 | 490 | |||
| XM_017003755.1 | XP_016859244.1 | 4 | 2074 | |||
| GenBank mRNA | AK127189.1 | - | 5 | 549 | ||
| AK302678.1 | - | 4 | 394 | |||
| AK302688.1 | - | 5 | 443 | |||
| BC031585.1 | - | 4 | 401 | |||
| BC047637.1 | - | 5 | 454 | |||
| BG723301.1 | - | 5 | 459 | |||
| BX649039.1 | - | 2 | 956 | |||