Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SGSM3 SIRNA Assays ›
          Gene Symbol:
          SGSM3
          Gene Name:
          small G protein signaling modulator 3
          Gene Aliases:
          MAP, RABGAP5, RUSC3, RUTBC3, RabGAP-5, rabGAPLP
          Chromosome Location:
          Chr.22: 40370530 - 40410289 on Build GRCh38
          UniGene ID:
          Hs.474914
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s226123

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SGSM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301849.1 NP_001288778.1 3 289
          NM_015705.5 NP_056520.2 4 372
          XM_005261572.2 XP_005261629.1 4 399
          XM_005261573.3 XP_005261630.1 4 404
          XM_005261574.4 XP_005261631.1 4 404
          XM_005261575.1 XP_005261632.1 3 267
          XM_011530158.1 XP_011528460.1 4 367
          XM_011530159.1 XP_011528461.1 4 349
          XM_011530160.1 XP_011528462.1 3 266
          XM_011530161.1 XP_011528463.1 4 349
          XM_011530162.2 XP_011528464.1 4 344
          XM_017028775.1 XP_016884264.1 3 253
          XM_017028776.1 XP_016884265.1 3 260
          XM_017028777.1 XP_016884266.1 3 260
          XM_017028778.1 XP_016884267.1 3 267
          XM_017028779.1 XP_016884268.1 3 266
          XM_017028780.1 XP_016884269.1 3 255
          GenBank mRNA AB196956.1 - 2 141
          AB275763.1 - 3 245
          AB449880.1 - 2 134
          AK295469.1 - 4 318
          AK297599.1 - 3 220
          AK301042.1 - 3 202
          AK309400.1 - 4 328
          AY308849.1 - 4 307
          BC008078.2 - 4 298 3027777
          BP283990.1 - 4 372
          BU585020.1 - 4 347
          CR456441.1 - 3 170

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          cell cycle arrest
          regulation of vesicle fusion
          regulation of Rab protein signal transduction
          Rap protein signal transduction
          plasma membrane to endosome transport
          activation of GTPase activity
          regulation of cilium assembly
          GTPase activator activity
          protein binding
          Rab GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2wn9f:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0