Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRICD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182563.3 | NP_872369.2 | 4, 5 | 666 | ||
XM_005255263.4 | XP_005255320.1 | 4, 5 | 2635 | |||
GenBank mRNA | AY597811.1 | - | 4, 5 | 431 | ||
BC039154.2 | - | 4, 5 | 608 | 4213860 |