Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARD8 SIRNA Assays ›
Gene Symbol:
CARD8
Gene Name:
caspase recruitment domain family member 8
Gene Aliases:
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location:
Chr.19: 48203148 - 48256269 on Build GRCh38
UniGene ID:
Hs.446146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22622
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184900.1
NP_001171829.1
8
1186
NM_001184901.1
NP_001171830.1
8
1133
NM_001184902.1
NP_001171831.1
11
1549
NM_001184903.1
NP_001171832.1
9
1283
NM_014959.3
NP_055774.2
9
1244
NR_033678.1
7
1103
NR_033679.1
7
1145
NR_033680.1
7
1103
XM_006723091.3
XP_006723154.1
10
1397
XM_006723092.3
XP_006723155.1
12
1935
XM_006723093.3
XP_006723156.1
10
1718
XM_006723094.3
XP_006723157.1
10
1377
XM_006723095.3
XP_006723158.1
10
1364
XM_006723096.3
XP_006723159.1
11
1786
XM_006723097.3
XP_006723160.1
9
1569
XM_006723098.3
XP_006723161.1
9
1356
XM_006723099.3
XP_006723162.1
8
1242
XM_006723101.2
XP_006723164.1
9
1259
XM_006723102.3
XP_006723165.1
8
1199
XM_006723104.3
XP_006723167.1
9
1256
XM_006723106.3
XP_006723169.1
10
1729
XM_006723109.3
XP_006723172.1
9
1256
XM_011526641.2
XP_011524943.1
13
2242
XM_011526642.2
XP_011524944.1
12
2137
XM_011526643.2
XP_011524945.1
11
1824
XM_011526644.2
XP_011524946.1
9
1409
XM_011526646.2
XP_011524948.1
10
1883
XM_011526647.2
XP_011524949.1
9
1772
XM_011526648.2
XP_011524950.1
10
1889
XM_011526650.2
XP_011524952.1
10
1380
XM_011526653.2
XP_011524955.1
10
1919
XM_017026482.1
XP_016881971.1
10
1722
XM_017026483.1
XP_016881972.1
10
1675
XM_017026484.1
XP_016881973.1
10
1683
XM_017026485.1
XP_016881974.1
10
1380
XM_017026486.1
XP_016881975.1
8
1239
XM_017026487.1
XP_016881976.1
7
1608
XM_017026488.1
XP_016881977.1
11
1824
XM_017026489.1
XP_016881978.1
10
1409
XM_017026490.1
XP_016881979.1
10
1380
XM_017026491.1
XP_016881980.1
10
1380
XM_017026492.1
XP_016881981.1
10
1691
XM_017026493.1
XP_016881982.1
9
1583
XM_017026494.1
XP_016881983.1
10
2197
XM_017026495.1
XP_016881984.1
9
1339
XM_017026496.1
XP_016881985.1
9
1259
XM_017026497.1
XP_016881986.1
8
1199
XM_017026498.1
XP_016881987.1
8
1196
XM_017026499.1
XP_016881988.1
7
2969
GenBank mRNA
AB023172.1
-
7
1138
AF322184.1
-
7
1138
AF331519.1
-
5
825
AF405558.1
-
5
825
AF511652.1
-
7
1138
AK297045.1
-
9
1269
AK297069.1
-
8
1123
AK302327.1
-
7
1073
AY026322.1
-
5
825
BX648568.1
-
9
3290
EU118120.1
-
8
1186
EU118121.1
-
7
1145
EU118122.1
-
5
986
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of interleukin-1 beta secretion
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
NACHT domain binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-89mxr:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0