Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARD8 SIRNA Assays ›
Gene Symbol:
CARD8
Gene Name:
caspase recruitment domain family member 8
Gene Aliases:
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location:
Chr.19: 48203148 - 48256269 on Build GRCh38
UniGene ID:
Hs.446146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22624
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184900.1
NP_001171829.1
5
556
NM_001184901.1
NP_001171830.1
5
503
NM_001184902.1
NP_001171831.1
8
919
NM_001184903.1
NP_001171832.1
6
653
NM_014959.3
NP_055774.2
6
614
NR_033678.1
4
476
NR_033679.1
4
515
NR_033680.1
4
476
XM_006723091.3
XP_006723154.1
7
767
XM_006723092.3
XP_006723155.1
9
1305
XM_006723093.3
XP_006723156.1
7
1088
XM_006723094.3
XP_006723157.1
7
750
XM_006723095.3
XP_006723158.1
7
734
XM_006723096.3
XP_006723159.1
8
1156
XM_006723097.3
XP_006723160.1
6
939
XM_006723098.3
XP_006723161.1
6
726
XM_006723099.3
XP_006723162.1
5
612
XM_006723101.2
XP_006723164.1
6
629
XM_006723102.3
XP_006723165.1
5
569
XM_006723104.3
XP_006723167.1
6
629
XM_006723106.3
XP_006723169.1
7
1099
XM_006723107.3
XP_006723170.1
7
750
XM_006723109.3
XP_006723172.1
6
629
XM_011526641.2
XP_011524943.1
10
1612
XM_011526642.2
XP_011524944.1
9
1507
XM_011526643.2
XP_011524945.1
8
1194
XM_011526644.2
XP_011524946.1
6
779
XM_011526646.2
XP_011524948.1
7
1253
XM_011526647.2
XP_011524949.1
6
1142
XM_011526648.2
XP_011524950.1
7
1262
XM_011526650.2
XP_011524952.1
7
750
XM_011526653.2
XP_011524955.1
7
1292
XM_017026482.1
XP_016881971.1
7
1095
XM_017026483.1
XP_016881972.1
7
1045
XM_017026484.1
XP_016881973.1
7
1056
XM_017026485.1
XP_016881974.1
7
750
XM_017026486.1
XP_016881975.1
5
612
XM_017026487.1
XP_016881976.1
4
978
XM_017026488.1
XP_016881977.1
8
1194
XM_017026489.1
XP_016881978.1
7
779
XM_017026490.1
XP_016881979.1
7
750
XM_017026491.1
XP_016881980.1
7
750
XM_017026492.1
XP_016881981.1
7
1061
XM_017026493.1
XP_016881982.1
6
953
XM_017026494.1
XP_016881983.1
7
1567
XM_017026495.1
XP_016881984.1
6
709
XM_017026496.1
XP_016881985.1
6
629
XM_017026497.1
XP_016881986.1
5
569
XM_017026498.1
XP_016881987.1
5
569
XM_017026499.1
XP_016881988.1
4
2342
GenBank mRNA
AB023172.1
-
4
511
AF322184.1
-
4
511
AF331519.1
-
2
198
AF405558.1
-
2
198
AF511652.1
-
4
511
AK297045.1
-
6
639
AK302327.1
-
4
443
AY026322.1
-
2
198
BX641091.1
-
1
426
BX648568.1
-
6
2660
EU118120.1
-
5
556
EU118121.1
-
4
515
EU118122.1
-
2
356
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of interleukin-1 beta secretion
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
NACHT domain binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0