Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFNA7 SIRNA Assays ›
          Gene Symbol:
          IFNA7 IFNA10 IFNA14 IFNA17 IFNA4
          Gene Name:
          interferon alpha 7
          interferon alpha 10
          interferon alpha 14
          interferon alpha 17
          interferon alpha 4
          Gene Aliases:
          IFN-alphaJ, IFNA-J
          IFN-alphaC
          IFN-alphaH, LEIF2H
          IFN-alphaI, IFNA, INFA, LEIF2C1
          IFN-alpha4a, INFA4
          Chromosome Location:
          Chr.9: 21201469 - 21202205 on Build GRCh38
          Chromosome Location:
          Chr.9: 21206181 - 21207143 on Build GRCh38
          Chromosome Location:
          Chr.9: 21239202 - 21239979 on Build GRCh38
          Chromosome Location:
          Chr.9: 21227243 - 21228222 on Build GRCh38
          Chromosome Location:
          Chr.9: 21186618 - 21187599 on Build GRCh38
          UniGene ID:
          Hs.282274
          Hs.282275
          Hs.93907
          Hs.282276
          Hs.1510
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s226337

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IFNA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021057.2 NP_066401.2 1 602
          GenBank mRNA BC074991.2 - 1 600 30915521
          BC074992.2 - 1 600 30915307
          BC113994.1 - 1 602 40084821
          BC114000.1 - 1 602 40084820
          M34913.1 - 1 602
          Gene Symbol: IFNA10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002171.2 NP_002162.1 1 608
          GenBank mRNA BC069409.1 - 1 608 7262186
          BC103972.2 - 1 608 40015081
          BC103973.1 - 1 608 40015082
          V00551.1 - 1 608
          Gene Symbol: IFNA14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002172.2 NP_002163.2 1 606
          GenBank mRNA BC074956.2 - 1 606 30915281
          BC104159.1 - 1 604 40029547
          BC104160.1 - 1 604 40029549
          V00542.1 - 1 618
          Gene Symbol: IFNA17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021268.2 NP_067091.1 1 611
          GenBank mRNA BC096732.1 - 1 604 40015059
          BC098355.1 - 1 604 40015056
          M11026.1 - 1 611
          M38289.1 - 1 704
          M71246.1 - 1 523
          Gene Symbol: IFNA4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021068.2 NP_066546.1 1 630
          GenBank mRNA BC074965.2 - 1 582 30915507
          BC074966.2 - 1 582 30915288
          BC113640.1 - 1 618 8322692
          BC113642.1 - 1 618 8322694
          M27318.1 - 1 702

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cell-cell signaling
          blood coagulation
          response to virus
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          innate immune response
          defense response to virus
          type I interferon signaling pathway
          regulation of type I interferon-mediated signaling pathway
          cytokine activity
          type I interferon receptor binding
          cytokine receptor binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zh286:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0