Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBNO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100122.1 | NP_001093592.1 | 20 | 2587 | ||
NM_014963.2 | NP_055778.2 | 23 | 2919 | |||
XM_011527804.2 | XP_011526106.1 | 23 | 2892 | |||
GenBank mRNA | AB023180.1 | - | 23 | 2896 | ||
AK074102.1 | - | 19 | 2379 | |||
AK125139.1 | - | 20 | 2534 | |||
AK292407.1 | - | 23 | 2919 |