Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP5-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012710.1 | NP_001012728.1 | 1 | 689 | ||
GenBank mRNA | AB126079.1 | - | 1 | 689 | ||
BC137550.1 | - | 1 | 689 | 9021550 | ||
BC137566.1 | - | 1 | 689 | 9021566 |