Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142550.1 | NP_001136022.1 | 12 | 2526 | ||
NM_001142551.1 | NP_001136023.1 | 12 | 2502 | |||
NM_014969.5 | NP_055784.3 | 12 | 2505 | |||
XM_006710460.3 | XP_006710523.1 | 12 | 2503 | |||
XM_011541027.2 | XP_011539329.1 | 12 | 2521 | |||
XM_011541028.2 | XP_011539330.1 | 12 | 2500 | |||
XM_011541029.1 | XP_011539331.1 | 12 | 2524 | |||
XM_011541030.1 | XP_011539332.1 | 12 | 2172 | |||
XM_011541031.1 | XP_011539333.1 | 10 | 1999 | |||
XM_017000696.1 | XP_016856185.1 | 12 | 2515 | |||
XM_017000697.1 | XP_016856186.1 | 12 | 2163 | |||
GenBank mRNA | AB020700.1 | - | 12 | 2349 | ||
AK074616.1 | - | 5 | 529 | |||
AK225781.1 | - | 12 | 2286 | |||
AK289789.1 | - | 12 | 2379 | |||
AK294998.1 | - | 9 | 1892 | |||
BC034964.2 | - | 12 | 2390 | 4823902 | ||
BC039254.1 | - | 11 | 2298 | 5287954 |