Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001891.3 | NP_001001891.2 | 12 | 1467 | ||
XM_011511263.2 | XP_011509565.1 | 12 | 1415 | |||
XM_011511267.2 | XP_011509569.1 | 12 | 1501 | |||
XM_017004229.1 | XP_016859718.1 | 12 | 1503 | |||
XM_017004230.1 | XP_016859719.1 | 11 | 1396 | |||
GenBank mRNA | AY617079.1 | - | 12 | 1468 |