Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016275.3 | NP_057359.2 | 2 | 323 | ||
GenBank mRNA | AF131856.1 | - | 2 | 278 | ||
AF195141.1 | - | 2 | 281 | |||
AK075006.1 | - | 2 | 296 | |||
AK300315.1 | - | 2 | 311 | |||
AY358095.1 | - | 2 | 287 | |||
AY358096.1 | - | 2 | 291 | |||
BC006012.1 | - | 2 | 314 | 4333083 | ||
BC008411.1 | - | 2 | 281 | 4132578 | ||
BC009556.1 | - | 2 | 255 | 3899430 | ||
BC009611.1 | - | 2 | 275 | 4093231 | ||
BC026350.1 | - | 2 | 303 | 4796883 | ||
BC036738.1 | - | 2 | 274 | 5540243 | ||
BC071699.1 | - | 2 | 266 | 5548565 | ||
CD707391.1 | - | 2 | 364 | |||
CR457154.1 | - | 1 | 65 |