Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DZANK1 SIRNA Assays ›
Gene Symbol:
DZANK1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location:
Chr.20: 18383367 - 18467185 on Build GRCh38
UniGene ID:
Hs.472225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s226825
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DZANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099407.1
NP_001092877.1
13
1482
XM_005260742.3
XP_005260799.1
13
1534
XM_005260743.3
XP_005260800.1
12
1388
XM_006723584.2
XP_006723647.1
11
1309
XM_011529268.2
XP_011527570.1
13
1652
XM_011529271.2
XP_011527573.1
13
2265
XM_011529272.2
XP_011527574.1
13
1648
XM_011529275.2
XP_011527577.1
12
2138
XM_011529277.2
XP_011527579.1
5
2379
XM_017027909.1
XP_016883398.1
13
1635
XM_017027910.1
XP_016883399.1
13
1518
XM_017027911.1
XP_016883400.1
13
1514
XM_017027912.1
XP_016883401.1
13
2221
XM_017027913.1
XP_016883402.1
13
2206
XM_017027914.1
XP_016883403.1
13
1460
XM_017027915.1
XP_016883404.1
13
1456
XM_017027916.1
XP_016883405.1
11
2020
XM_017027917.1
XP_016883406.1
14
2315
XM_017027918.1
XP_016883407.1
14
1569
XM_017027919.1
XP_016883408.1
14
1565
XM_017027920.1
XP_016883409.1
12
1524
XM_017027921.1
XP_016883410.1
6
686
GenBank mRNA
AK001462.1
-
7
669
AK055544.1
-
12
1277
AK131451.1
-
14
2121
AK295073.1
-
13
1392
AK299746.1
-
13
1405
BC036231.1
-
13
2730
BC130341.1
-
12
1310
40146372
BC144153.1
-
12
1310
9052673
CR749327.1
-
14
1534
DQ104738.1
-
4
403
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitophagy in response to mitochondrial depolarization
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-wfnwd:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0