Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007267.2 | NP_001007268.1 | 14 | 2467 | ||
NM_001195641.1 | NP_001182570.1 | 14 | 2467 | |||
NM_007366.4 | NP_031392.3 | 14 | 2467 | |||
XM_005246392.3 | XP_005246449.1 | 14 | 2355 | |||
XM_011510820.2 | XP_011509122.1 | 14 | 2355 | |||
XM_017003598.1 | XP_016859087.1 | 14 | 2355 | |||
XM_017003599.1 | XP_016859088.1 | 14 | 2355 | |||
XM_017003600.1 | XP_016859089.1 | 4 | 578 | |||
XM_017003601.1 | XP_016859090.1 | 14 | 2754 | |||
XM_017003602.1 | XP_016859091.1 | 3 | 304 | |||
GenBank mRNA | BC140823.1 | - | 14 | 2277 | 9021691 | |
BC144631.1 | - | 14 | 2277 | 9053162 | ||
BX485732.1 | - | 2 | 119 | |||
D30780.1 | - | 5 | 662 | |||
U17033.1 | - | 14 | 2467 | |||
U17034.1 | - | 14 | 2467 |