Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1456 SIRNA Assays ›
Gene Symbol:
KIAA1456
Gene Name:
KIAA1456
Gene Aliases:
C8orf79, TRM9L
Chromosome Location:
Chr.8: 12945674 - 13029777 on Build GRCh38
UniGene ID:
Hs.202521
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s226922
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1456
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099677.1
NP_001093147.1
2
1361
NM_020844.2
NP_065895.2
5
1651
XM_005273584.4
XP_005273641.1
5
1662
XM_005273585.4
XP_005273642.1
6
1673
XM_005273586.4
XP_005273643.1
6
1707
XM_005273587.4
XP_005273644.1
5
1895
XM_005273588.4
XP_005273645.1
6
1819
XM_005273590.4
XP_005273647.1
4
1351
XM_005273591.4
XP_005273648.1
5
1835
XM_011544596.2
XP_011542898.1
5
2122
XM_011544597.2
XP_011542899.1
5
1474
XM_011544598.2
XP_011542900.1
5
1743
XM_011544600.2
XP_011542902.1
5
1481
XM_011544601.2
XP_011542903.1
5
1604
XM_017013705.1
XP_016869194.1
5
1512
XM_017013706.1
XP_016869195.1
5
1733
XM_017013707.1
XP_016869196.1
6
1878
XM_017013708.1
XP_016869197.1
5
1977
XM_017013709.1
XP_016869198.1
6
1862
XM_017013710.1
XP_016869199.1
7
1821
XM_017013711.1
XP_016869200.1
6
1656
XM_017013712.1
XP_016869201.1
6
2033
XM_017013713.1
XP_016869202.1
5
1777
XM_017013714.1
XP_016869203.1
4
1579
XM_017013715.1
XP_016869204.1
5
2452
GenBank mRNA
AB040889.1
-
2
1425
AK307825.1
-
2
1677
BC016633.2
-
2
1004
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
tRNA wobble uridine modification
tRNA modification
tRNA methylation
oxidation-reduction process
tRNA binding
tRNA methyltransferase activity
ferrous iron binding
tRNA (uracil) methyltransferase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0