Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36B SIRNA Assays ›
          Gene Symbol:
          ANKRD36B ANKRD36
          Gene Name:
          ankyrin repeat domain 36B
          ankyrin repeat domain 36
          Gene Aliases:
          KIAA1641
          UNQ2430
          Chromosome Location:
          Chr.2: 97492663 - 97590245 on Build GRCh38
          Chromosome Location:
          Chr.2: 97113186 - 97264521 on Build GRCh38
          UniGene ID:
          Hs.532921
          Hs.541894
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s226927

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_025190.3 NP_079466.3 24, 25 1968
          XM_005263995.4 XP_005264052.1 24, 25 2248
          XM_006712661.1 XP_006712724.1 24, 25 2248
          XM_011511555.1 XP_011509857.1 24, 25 2248
          XM_011511556.1 XP_011509858.1 24, 25 2248
          XM_011511557.2 XP_011509859.1 24, 25 2248
          XM_011511558.1 XP_011509860.1 24, 25 2248
          XM_017004578.1 XP_016860067.1 24, 25 2248
          XM_017004579.1 XP_016860068.1 24, 25 2248
          XM_017004580.1 XP_016860069.1 22, 23 2146
          XM_017004581.1 XP_016860070.1 22, 23 2146
          XM_017004583.1 XP_016860072.1 23, 24 2110
          XM_017004585.1 XP_016860074.1 22, 23 2003
          XM_017004586.1 XP_016860075.1 19, 20 989
          XM_017004587.1 XP_016860076.1 23, 24 2180
          XM_017004588.1 XP_016860077.1 22, 23 2042
          XM_017004589.1 XP_016860078.1 21, 22 1935
          XM_017004590.1 XP_016860079.1 20, 21 1862
          XM_017004591.1 XP_016860080.1 6, 7 1966
          XM_017004592.1 XP_016860081.1 25, 26 2292
          XM_017004593.1 XP_016860082.1 23, 24 2098
          XM_017004594.1 XP_016860083.1 22, 23 1991
          XM_017004595.1 XP_016860084.1 23, 24 2047
          GenBank mRNA BC031883.2 - 7, 8 387
          BC044844.2 - 7, 8 387
          Gene Symbol: ANKRD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164315.1 NP_001157787.1 58, 59 3720
          XM_006712514.3 XP_006712577.1 58, 59 4030
          XM_011511130.1 XP_011509432.1 58, 59 4030
          XM_011511131.1 XP_011509433.1 58, 59 4030
          XM_017004009.1 XP_016859498.1 58, 59 4030
          XM_017004010.1 XP_016859499.1 58, 59 4030
          XM_017004011.1 XP_016859500.1 56, 57 3928
          XM_017004012.1 XP_016859501.1 56, 57 3928
          XM_017004013.1 XP_016859502.1 56, 57 3928
          XM_017004014.1 XP_016859503.1 56, 57 3928
          XM_017004015.1 XP_016859504.1 56, 57 3928
          XM_017004016.1 XP_016859505.1 56, 57 3928
          XM_017004017.1 XP_016859506.1 56, 57 3928
          XM_017004018.1 XP_016859507.1 56, 57 3928
          XM_017004019.1 XP_016859508.1 56, 57 3928
          XM_017004020.1 XP_016859509.1 56, 57 3928
          XM_017004021.1 XP_016859510.1 56, 57 3928
          XM_017004022.1 XP_016859511.1 56, 57 3928
          XM_017004024.1 XP_016859513.1 57, 58 3892
          XM_017004025.1 XP_016859514.1 54, 55 3826
          XM_017004026.1 XP_016859515.1 54, 55 3826
          XM_017004027.1 XP_016859516.1 54, 55 3826
          XM_017004028.1 XP_016859517.1 54, 55 3826
          XM_017004029.1 XP_016859518.1 54, 55 3826
          XM_017004030.1 XP_016859519.1 54, 55 3826
          XM_017004032.1 XP_016859521.1 54, 55 3826
          XM_017004033.1 XP_016859522.1 54, 55 3826
          XM_017004034.1 XP_016859523.1 54, 55 3826
          XM_017004035.1 XP_016859524.1 54, 55 3826
          XM_017004036.1 XP_016859525.1 54, 55 3826
          XM_017004038.1 XP_016859527.1 54, 55 3826
          XM_017004039.1 XP_016859528.1 52, 53 3724
          XM_017004040.1 XP_016859529.1 52, 53 3724
          XM_017004041.1 XP_016859530.1 52, 53 3724
          XM_017004042.1 XP_016859531.1 52, 53 3724
          XM_017004044.1 XP_016859533.1 50, 51 3622
          XM_017004045.1 XP_016859534.1 50, 51 3622
          XM_017004046.1 XP_016859535.1 56, 57 3785
          XM_017004047.1 XP_016859536.1 55, 56 3717
          XM_017004048.1 XP_016859537.1 54, 55 3644
          XM_017004049.1 XP_016859538.1 56, 57 3819
          XM_017004050.1 XP_016859539.1 41, 42 2223
          XM_017004051.1 XP_016859540.1 39, 40 2437

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0