siRNA Details
Gene Symbol: SIRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193286.1 | NP_001180215.1 | 9 | 956 | ||
NM_012237.3 | NP_036369.2 | 10 | 964 | |||
NM_030593.2 | NP_085096.1 | 9 | 956 | |||
NR_034146.1 | 8 | 887 | ||||
XM_006723111.1 | XP_006723174.1 | 11 | 912 | |||
XM_011526654.1 | XP_011524956.1 | 9 | 849 | |||
XM_011526655.1 | XP_011524957.1 | 7 | 664 | |||
XM_017026500.1 | XP_016881989.1 | 9 | 888 | |||
GenBank mRNA | AB009274.1 | - | 9 | 677 | ||
AF083107.2 | - | 10 | 862 | |||
AF095714.1 | - | 9 | 735 | |||
AF131800.1 | - | 8 | 611 | |||
AF160214.1 | - | 9 | 716 | |||
AK025876.1 | - | 9 | 742 | |||
AK054642.1 | - | 10 | 795 | |||
AK131390.1 | - | 4 | 1914 | |||
AK225824.1 | - | 9 | 739 | |||
AK290716.1 | - | 8 | 887 | |||
AK314492.1 | - | 10 | 718 | |||
AY030277.1 | - | 9 | 717 | |||
BC003012.1 | - | 9 | 807 | 2820929 | ||
BC003547.1 | - | 9 | 807 | 3536728 | ||
KF032391.1 | - | 7 | 664 |

More Information
Panther Classification:
Molecular Function -





