Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SIRT2 SIRNA Assays ›
          Gene Symbol:
          SIRT2
          Gene Name:
          sirtuin 2
          Gene Aliases:
          SIR2, SIR2L, SIR2L2
          Chromosome Location:
          Chr.19: 38878555 - 38899862 on Build GRCh38
          UniGene ID:
          Hs.466693
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22708

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SIRT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012237.3 NP_036369.2 13 1159
          NM_030593.2 NP_085096.1 12 1151
          NR_034146.1 11 1082
          XM_006723111.1 XP_006723174.1 14 1107
          XM_011526654.1 XP_011524956.1 12 1044
          XM_011526655.1 XP_011524957.1 10 859
          XM_017026500.1 XP_016881989.1 12 1083
          GenBank mRNA AB009274.1 - 12 872
          AF083107.2 - 13 1057
          AF095714.1 - 12 930
          AF160214.1 - 12 911
          AJ505014.1 - 10 1628
          AK025876.1 - 12 937
          AK054642.1 - 13 990
          AK131390.1 - 5 2587
          AK225824.1 - 12 934
          AK290716.1 - 11 1082
          AK314492.1 - 13 913
          AY030277.1 - 12 912
          BC003012.1 - 12 1002 2820929
          BC003547.1 - 12 1002 3536728
          KF032391.1 - 10 859

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          chromatin silencing at rDNA
          chromatin silencing
          chromatin silencing at telomere
          transcription, DNA-templated
          protein ADP-ribosylation
          protein deacetylation
          autophagy
          mitotic nuclear division
          regulation of exit from mitosis
          negative regulation of cell proliferation
          negative regulation of autophagy
          negative regulation of peptidyl-threonine phosphorylation
          phosphatidylinositol 3-kinase signaling
          gene silencing
          histone deacetylation
          substantia nigra development
          myelination in peripheral nervous system
          regulation of myelination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          cellular response to oxidative stress
          peptidyl-lysine deacetylation
          cellular response to hepatocyte growth factor stimulus
          negative regulation of protein catabolic process
          regulation of phosphorylation
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of DNA binding
          protein kinase B signaling
          cellular lipid catabolic process
          innate immune response
          negative regulation of fat cell differentiation
          positive regulation of meiotic nuclear division
          negative regulation of striated muscle tissue development
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          hepatocyte growth factor receptor signaling pathway
          negative regulation of oligodendrocyte differentiation
          cell division
          meiotic cell cycle
          regulation of cell cycle
          response to redox state
          positive regulation of cell division
          positive regulation of attachment of spindle microtubules to kinetochore
          negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
          cellular response to caloric restriction
          negative regulation of oligodendrocyte progenitor proliferation
          histone H3 deacetylation
          histone H4 deacetylation
          cellular response to molecule of bacterial origin
          cellular response to hypoxia
          cellular response to epinephrine stimulus
          tubulin deacetylation
          positive regulation of execution phase of apoptosis
          positive regulation of oocyte maturation
          negative regulation of NLRP3 inflammasome complex assembly
          negative regulation of defense response to bacterium
          ripoptosome assembly involved in necroptotic process
          negative regulation of reactive oxygen species metabolic process
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process involved in cellular response to hypoxia
          chromatin binding
          NAD+ ADP-ribosyltransferase activity
          histone deacetylase activity
          protein binding
          transcription factor binding
          zinc ion binding
          NAD-dependent histone deacetylase activity
          protein deacetylase activity
          NAD-dependent protein deacetylase activity
          histone acetyltransferase binding
          histone deacetylase binding
          tubulin deacetylase activity
          ubiquitin binding
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          beta-tubulin binding
          NAD+ binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gh6bl:80/100.66.76.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0