Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198542.1 | NP_001185471.1 | 19 | 2842 | ||
NM_001198543.1 | NP_001185472.1 | 19 | 2812 | |||
NM_001270974.2 | NP_001257903.1 | 19 | 2910 | |||
NM_017558.4 | NP_060028.2 | 19 | 2910 | |||
XM_006721206.2 | XP_006721269.1 | 19 | 2804 | |||
XM_011523146.2 | XP_011521448.1 | 20 | 2939 | |||
XM_011523147.1 | XP_011521449.1 | 20 | 2906 | |||
XM_011523148.1 | XP_011521450.1 | 20 | 2979 | |||
XM_011523151.1 | XP_011521453.1 | 19 | 2836 | |||
XM_017023346.1 | XP_016878835.1 | 19 | 3082 | |||
XM_017023347.1 | XP_016878836.1 | 7 | 1321 | |||
XM_017023348.1 | XP_016878837.1 | 7 | 1061 | |||
GenBank mRNA | AK022933.1 | - | 19 | 2844 | ||
AK057467.1 | - | 3 | 1214 | |||
AK299016.1 | - | 19 | 2799 | |||
AK299348.1 | - | 19 | 2842 | |||
AK308296.1 | - | 17 | 2429 | |||
AL122038.1 | - | 4 | 585 | |||
AL133042.1 | - | 4 | 585 |
Gene Symbol: HYDIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103556.1 | 14 | 2090 | |||
GenBank mRNA | AL833826.1 | - | 16 | 2286 | ||
AL834340.1 | - | 11 | 1580 |