Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POTEJ SIRNA Assays ›
          Gene Symbol:
          POTEJ LOC100288966 LOC102723502 POTEB POTEB2 POTEB3 POTEC POTED POTEE POTEF POTEG POTEH POTEI POTEM
          Gene Name:
          POTE ankyrin domain family member J
          POTE ankyrin domain family member D-like
          POTE ankyrin domain family member B-like
          POTE ankyrin domain family member B
          POTE ankyrin domain family member B2
          POTE ankyrin domain family member B3
          POTE ankyrin domain family member C
          POTE ankyrin domain family member D
          POTE ankyrin domain family member E
          POTE ankyrin domain family member F
          POTE ankyrin domain family member G
          POTE ankyrin domain family member H
          POTE ankyrin domain family member I
          POTE ankyrin domain family member M
          Gene Aliases:
          POTE2beta
          -
          -
          A26B1, CT104.5, POTE-15, POTE15
          -
          POTE-15, POTEB
          A26B2, CT104.6, POTE-18, POTE18
          A26B3, ANKRD21, CT104.1, POTE, POTE-21, POTE21
          A26C1, A26C1A, CT104.2, POTE-2, POTE2, POTE2gamma
          A26C1B, ACTB, POTE2alpha, POTEACTIN
          A26C2, ACTBL1, CT104.4, POTE-14, POTE14, POTE14alpha, POTE22
          A26C3, ACTBL1, CT104.7, POTE22
          POTE2beta
          ACT, P704P, POTE14beta
          Chromosome Location:
          Chr.2: 130611077 - 130658448 on Build GRCh38
          Chromosome Location:
          Chr.Un|NT_167208: 108007 - 139435 on Build GRCh38
          Chromosome Location:
          Chr.15|NT_187382: 132942 - 167347 on Build GRCh38
          Chromosome Location:
          Chr.15: 21846329 - 21877393 on Build GRCh38
          Chromosome Location:
          Chr.15: 20835372 - 20866428 on Build GRCh38
          Chromosome Location:
          Chr.15: 21408964 - 21440429 on Build GRCh38
          Chromosome Location:
          Chr.18: 14507339 - 14543600 on Build GRCh38
          Chromosome Location:
          Chr.21: 13610081 - 13641585 on Build GRCh38
          Chromosome Location:
          Chr.2: 131209580 - 131265254 on Build GRCh38
          Chromosome Location:
          Chr.2: 130073535 - 130129222 on Build GRCh38
          Chromosome Location:
          Chr.14: 19402764 - 19434341 on Build GRCh38
          Chromosome Location:
          Chr.22: 15690026 - 15721631 on Build GRCh38
          Chromosome Location:
          Chr.2: 130462245 - 131626428 on Build GRCh38
          Chromosome Location:
          Chr.14: 18967434 - 19003752 on Build GRCh38
          UniGene ID:
          Hs.558829
          Hs.442712
          -
          Hs.722659
          Hs.722659
          Hs.722659
          Hs.454726
          Hs.742365
          Hs.686146
          Hs.580547
          Hs.711826
          Hs.581075
          Hs.558782
          Hs.654289
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s227419

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POTEJ
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277083.1 NP_001264012.1 5 897
          XM_011511683.2 XP_011509985.1 5 1069
          XM_017004741.1 XP_016860230.1 6 1042
          Gene Symbol: LOC100288966
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257362.1 NP_001244291.1 5 1060
          XM_006709930.3 XP_006709993.1 5 1156
          XM_011545655.2 XP_011543957.1 5 1156
          XM_011545656.2 XP_011543958.1 5 1156
          GenBank mRNA AY172978.1 - 5 1060
          Gene Symbol: LOC102723502
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006724900.3 XP_006724963.1 5 1155
          XM_006724901.3 XP_006724964.1 5 1155
          XM_011546199.2 XP_011544501.1 5 1155
          XM_011546200.2 XP_011544502.1 5 1155
          Gene Symbol: POTEB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277304.1 NP_001264233.1 5 930
          XM_006720355.3 XP_006720418.1 5 1044
          XM_011543794.2 XP_011542096.1 5 1044
          XM_017021858.1 XP_016877347.1 5 1044
          Gene Symbol: POTEB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277303.1 NP_001264232.1 5 930
          XM_006720354.3 XP_006720417.1 5 1044
          XM_011543793.2 XP_011542095.1 5 1044
          XM_017021853.1 XP_016877342.1 5 1044
          Gene Symbol: POTEB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_207355.3 NP_997238.2 5 1379
          NR_110752.1 5 1379
          XM_006720358.2 XP_006720421.1 5 1426
          XM_011543795.2 XP_011542097.1 5 1426
          XM_011543796.2 XP_011542098.1 5 1426
          XM_011543797.2 XP_011542099.1 5 1426
          GenBank mRNA AY465170.1 - 6 949
          AY465173.1 - 5 1056
          BC066981.1 - 5 1379 4824630
          BC101005.1 - 5 930 40006483
          BC101006.1 - 5 1041 40006485
          BC127123.1 - 5 1041 40006490
          Gene Symbol: POTEC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001137671.1 NP_001131143.1 5 1462
          NR_134878.1 5 1462
          GenBank mRNA AK307658.1 - 5 1462
          BX649118.1 - 5 1448
          Gene Symbol: POTED
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_174981.3 NP_778146.2 5 1060
          XM_006723997.3 XP_006724060.1 5 1156
          XM_011529550.2 XP_011527852.1 5 1156
          Gene Symbol: POTEE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083538.1 NP_001077007.1 5 1060
          XM_011511216.2 XP_011509518.1 5 1138
          XM_017004159.1 XP_016859648.1 5 1169
          XM_017004161.1 XP_016859650.1 7 1497
          GenBank mRNA AY462868.1 - 5 1060
          AY462871.1 - 5 1060
          AY462872.1 - 5 1090
          AY462873.1 - 5 1060
          Gene Symbol: POTEF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099771.2 NP_001093241.1 7 1408
          XM_017004833.1 XP_016860322.1 5 1183
          GenBank mRNA EF523384.1 - 7 1410
          Gene Symbol: POTEG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005356.2 NP_001005356.1 5 1060
          NR_027480.1 5 1060
          GenBank mRNA AY462874.1 - 5 1060
          AY465171.1 - 7 1318
          AY465172.1 - 6 1060
          BC127623.1 - 5 1040
          Gene Symbol: POTEH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136213.1 NP_001129685.1 5 1171
          GenBank mRNA AY466021.1 - 6 1370
          Gene Symbol: POTEI
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277406.1 NP_001264335.1 5 1008
          XM_011511670.2 XP_011509972.1 5 1180
          XM_017004732.1 XP_016860221.1 6 1159
          Gene Symbol: POTEM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145442.1 NP_001138914.1 5 1060

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          substantia nigra development
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0