Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007349.3 | NP_031375.3 | 7 | 1148 | ||
XM_005249539.1 | XP_005249596.1 | 8 | 1287 | |||
XM_011515982.2 | XP_011514284.1 | 7 | 1262 | |||
XM_011515983.1 | XP_011514285.1 | 7 | 1148 | |||
XM_017011881.1 | XP_016867370.1 | 8 | 2460 | |||
GenBank mRNA | AK123044.1 | - | 3 | 1193 | ||
AK123600.1 | - | 8 | 1287 | |||
AK293744.1 | - | 5 | 980 | |||
AK307417.1 | - | 7 | 1151 | |||
BC033781.1 | - | 2 | 389 | 4509264 | ||
BX640616.1 | - | 7 | 3046 | |||
U80735.1 | - | 2 | 503 |