Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NT5C2 SIRNA Assays ›
Gene Symbol:
NT5C2
Gene Name:
5'-nucleotidase, cytosolic II
Gene Aliases:
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location:
Chr.10: 103088017 - 103193306 on Build GRCh38
UniGene ID:
Hs.97439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22756
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NT5C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134373.2
NP_001127845.1
17
1468
NM_012229.4
NP_036361.1
18
1566
XM_005269632.4
XP_005269689.1
18
1477
XM_005269633.4
XP_005269690.1
19
1570
XM_005269634.4
XP_005269691.1
19
1602
XM_005269635.4
XP_005269692.1
18
1792
XM_005269636.4
XP_005269693.1
18
1578
XM_005269637.4
XP_005269694.1
17
2296
XM_005269638.4
XP_005269695.1
17
3597
XM_005269639.4
XP_005269696.1
16
1321
XM_005269640.4
XP_005269697.1
19
1593
XM_005269641.4
XP_005269698.1
18
1495
XM_005269642.4
XP_005269699.1
17
1420
XM_005269643.4
XP_005269700.1
16
1360
XM_005269644.4
XP_005269701.1
20
1617
XM_005269645.4
XP_005269702.1
19
1519
XM_005269646.4
XP_005269703.1
18
1445
XM_006717721.3
XP_006717784.1
18
1519
XM_006717723.3
XP_006717786.1
16
1316
XM_011539537.2
XP_011537839.1
18
1431
XM_017015947.1
XP_016871436.1
16
1346
XM_017015948.1
XP_016871437.1
19
1702
XM_017015949.1
XP_016871438.1
19
1687
XM_017015950.1
XP_016871439.1
17
1403
XM_017015951.1
XP_016871440.1
16
1329
XM_017015952.1
XP_016871441.1
18
1626
XM_017015953.1
XP_016871442.1
19
2021
XM_017015954.1
XP_016871443.1
18
1960
XM_017015955.1
XP_016871444.1
18
1647
XM_017015956.1
XP_016871445.1
18
1853
XM_017015957.1
XP_016871446.1
20
1711
XM_017015958.1
XP_016871447.1
21
1830
XM_017015959.1
XP_016871448.1
17
1421
XM_017015960.1
XP_016871449.1
18
1384
XM_017015961.1
XP_016871450.1
18
1779
XM_017015962.1
XP_016871451.1
18
1613
XM_017015963.1
XP_016871452.1
18
1717
XM_017015964.1
XP_016871453.1
18
1696
XM_017015965.1
XP_016871454.1
17
1573
XM_017015966.1
XP_016871455.1
17
1573
XM_017015967.1
XP_016871456.1
17
1508
XM_017015968.1
XP_016871457.1
18
1501
XM_017015969.1
XP_016871458.1
18
1595
XM_017015970.1
XP_016871459.1
16
2238
XM_017015971.1
XP_016871460.1
15
1282
XM_017015972.1
XP_016871461.1
19
1536
XM_017015973.1
XP_016871462.1
20
1560
XM_017015974.1
XP_016871463.1
19
1560
XM_017015975.1
XP_016871464.1
17
1401
XM_017015976.1
XP_016871465.1
17
1407
GenBank mRNA
AK127670.1
-
18
1557
AK291667.1
-
17
1434
AK295593.1
-
16
1610
AK312517.1
-
17
1434
BC001595.1
-
17
1517
3543103
BX648078.1
-
17
5468
D38524.1
-
17
1456
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00366992_m1
Panther Classification:
Molecular Function -
nucleotide phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide catabolic process
phosphorylation
dephosphorylation
drug metabolic process
IMP metabolic process
adenosine metabolic process
nucleotide binding
protein binding
5'-nucleotidase activity
metal ion binding
nucleoside phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0