Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHRNA7 SIRNA Assays ›
          Gene Symbol:
          CHRNA7 CHRFAM7A
          Gene Name:
          cholinergic receptor nicotinic alpha 7 subunit
          CHRNA7 (exons 5-10) and FAM7A (exons A-E) fusion
          Gene Aliases:
          CHRNA7-2, NACHRA7
          CHRNA7, CHRNA7-DR1, D-10
          Chromosome Location:
          Chr.15: 32030462 - 32172521 on Build GRCh38
          Chromosome Location:
          Chr.15: 30357770 - 30393849 on Build GRCh38
          UniGene ID:
          Hs.511772
          Hs.510853
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s228143

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHRNA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000746.5 NP_000737.1 10 1264
          NM_001190455.2 NP_001177384.1 10 1351
          NR_046324.1 8 1074
          XM_011521176.2 XP_011519478.2 9 1119
          XM_011521177.2 XP_011519479.1 7 1204
          XM_011521178.2 XP_011519480.1 9 1154
          XM_017021882.1 XP_016877371.1 11 1329
          XM_017021883.1 XP_016877372.1 11 1326
          XM_017021884.1 XP_016877373.1 9 1175
          GenBank mRNA AF385585.1 - 10 1166
          AJ586911.1 - 10 1169
          AK292069.1 - 10 1234
          AK294229.1 - 10 1346
          BC037571.1 - 8 1001 5245812
          L25827.1 - 9 1129
          U40583.1 - 10 1159
          U62436.1 - 10 1224
          X70297.1 - 10 1255
          Y08420.1 - 10 1152
          Z23141.1 - 9 1083
          Gene Symbol: CHRFAM7A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_139320.1 NP_647536.1 10 1448
          NM_148911.1 NP_683709.1 9 1384
          XM_005254750.2 XP_005254807.1 11 1617
          XM_011522153.1 XP_011520455.1 11 1142
          XM_011522155.2 XP_011520457.1 9 1333
          GenBank mRNA AF036903.1 - 8 894
          AF037646.1 - 7 1018
          AK292984.1 - 10 1215
          BC101345.1 - 11 1321 40025185
          BC101346.2 - 10 1147 40025186
          BC101347.2 - 10 1147 40025187
          BC101348.2 - 10 1147 40025188

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          response to hypoxia
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          signal transduction
          synaptic transmission, cholinergic
          memory
          positive regulation of cell proliferation
          negative regulation of tumor necrosis factor production
          ion transmembrane transport
          response to nicotine
          positive regulation of angiogenesis
          cognition
          cation transmembrane transport
          beta-amyloid binding
          acetylcholine-activated cation-selective channel activity
          ligand-gated ion channel activity
          acetylcholine receptor activity
          toxic substance binding
          chloride channel regulator activity
          acetylcholine-gated cation channel activity
          acetylcholine binding
          protein homodimerization activity
          extracellular ligand-gated ion channel activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0