Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003592.2 | NP_003583.2 | 4 | 691 | ||
XM_005250060.4 | XP_005250117.1 | 4 | 727 | |||
XM_011516629.2 | XP_011514931.1 | 4 | 683 | |||
XM_011516630.2 | XP_011514932.1 | 4 | 584 | |||
XM_011516631.2 | XP_011514933.1 | 4 | 712 | |||
XM_011516632.2 | XP_011514934.1 | 4 | 610 | |||
XM_017012723.1 | XP_016868212.1 | 4 | 611 | |||
GenBank mRNA | AF062536.1 | - | 3 | 465 | ||
AK096163.1 | - | 5 | 579 | |||
AK315023.1 | - | 4 | 867 | |||
BC125119.1 | - | 4 | 604 | 40118923 | ||
BC125120.1 | - | 4 | 604 | 40118927 | ||
BG391038.1 | - | 708 | ||||
BX537409.1 | - | 4 | 621 | |||
U58087.1 | - | 4 | 393 |