Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310338.1 | NP_001297267.1 | 5 | 1140 | ||
NM_052865.3 | NP_443097.1 | 4 | 1095 | |||
XM_005260867.2 | XP_005260924.1 | 5 | 1362 | |||
XM_011529394.1 | XP_011527696.1 | 5 | 977 | |||
XM_011529395.1 | XP_011527697.1 | 4 | 855 | |||
XM_017028127.1 | XP_016883616.1 | 4 | 1317 | |||
XM_017028128.1 | XP_016883617.1 | 4 | 1191 | |||
GenBank mRNA | AF306685.1 | - | 4 | 904 | ||
AK027503.1 | - | 4 | 899 | |||
AK309563.1 | - | 2 | 438 | |||
AK309995.1 | - | 2 | 366 | |||
AK310282.1 | - | 3 | 411 | |||
AK315532.1 | - | 4 | 931 | |||
BC016869.1 | - | 4 | 867 | 3850853 |