Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFAND4 SIRNA Assays ›
Gene Symbol:
ZFAND4
Gene Name:
zinc finger AN1-type containing 4
Gene Aliases:
ANUBL1
Chromosome Location:
Chr.10: 45614567 - 45672962 on Build GRCh38
UniGene ID:
Hs.659717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s228496
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFAND4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128324.2
NP_001121796.1
2
369
NM_001282905.1
NP_001269834.1
2
717
NM_001282906.1
NP_001269835.1
2
369
NM_174890.3
NP_777550.2
2
717
NR_104256.1
2
369
XM_005271842.2
XP_005271899.1
2
318
XM_005271843.2
XP_005271900.1
2
485
XM_011540366.2
XP_011538668.1
2
457
XM_017016931.1
XP_016872420.1
2
459
XM_017016932.1
XP_016872421.1
2
461
XM_017016933.1
XP_016872422.1
2
328
XM_017016934.1
XP_016872423.1
2
381
XM_017016935.1
XP_016872424.1
1
131
XM_017016936.1
XP_016872425.1
2
615
XM_017016937.1
XP_016872426.1
2
463
XM_017016938.1
XP_016872427.1
2
775
XM_017016939.1
XP_016872428.1
2
346
XM_017016940.1
XP_016872429.1
2
775
XM_017016941.1
XP_016872430.1
2
318
XM_017016942.1
XP_016872431.1
2
485
XM_017016943.1
XP_016872432.1
2
761
XM_017016944.1
XP_016872433.1
2
318
XM_017016946.1
XP_016872435.1
2
328
GenBank mRNA
AF311324.1
-
2
333
AK090704.1
-
3
543
AK097924.1
-
2
336
AK292420.1
-
2
318
AK292469.1
-
2
318
AK314395.1
-
2
329
BC045587.1
-
2
369
5269483
BC048968.1
-
2
259
5272133
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0