Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMCC1 SIRNA Assays ›
          Gene Symbol:
          TMCC1
          Gene Name:
          transmembrane and coiled-coil domain family 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 129647792 - 129893605 on Build GRCh38
          UniGene ID:
          Hs.477547
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22852

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017395.3 NP_001017395.2 4 1609
          NM_001128224.2 NP_001121696.1 4 1010
          NR_033361.1 2 1155
          XM_006713542.3 XP_006713605.1 5 1722
          XM_006713543.3 XP_006713606.1 5 1719
          XM_006713544.1 XP_006713607.1 5 1847
          XM_006713550.3 XP_006713613.1 5 1280
          XM_006713552.3 XP_006713615.1 2 902
          XM_006713553.3 XP_006713616.1 2 2194
          XM_011512572.2 XP_011510874.1 6 1882
          XM_011512573.2 XP_011510875.1 5 2461
          XM_011512576.2 XP_011510878.1 5 1715
          XM_011512579.2 XP_011510881.1 4 1470
          XM_011512580.2 XP_011510882.1 7 1607
          XM_011512581.2 XP_011510883.1 6 1444
          XM_011512582.1 XP_011510884.1 2 1236
          XM_011512583.2 XP_011510885.1 4 1013
          XM_011512584.2 XP_011510886.1 6 1332
          XM_017005931.1 XP_016861420.1 5 1630
          XM_017005932.1 XP_016861421.1 6 1869
          XM_017005933.1 XP_016861422.1 4 1666
          XM_017005934.1 XP_016861423.1 8 1680
          XM_017005935.1 XP_016861424.1 9 1769
          XM_017005936.1 XP_016861425.1 6 1443
          XM_017005937.1 XP_016861426.1 6 1377
          XM_017005938.1 XP_016861427.1 8 1743
          XM_017005939.1 XP_016861428.1 7 1637
          XM_017005940.1 XP_016861429.1 4 1015
          XM_017005941.1 XP_016861430.1 4 1141
          GenBank mRNA AB018322.1 - 1 176
          AK124671.1 - 2 1156
          AK291448.1 - 4 1428
          AK293408.1 - 2 692
          BC030619.2 - 2 1073
          BC039859.1 - 4 1516 5527623
          BC047517.1 - 2 1073
          BX640712.1 - 2 663
          CR749206.1 - 2 1135

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0