Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMCC1 SIRNA Assays ›
          Gene Symbol:
          TMCC1
          Gene Name:
          transmembrane and coiled-coil domain family 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 129647792 - 129893605 on Build GRCh38
          UniGene ID:
          Hs.477547
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22853

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017395.3 NP_001017395.2 4, 5 1942
          NM_001128224.2 NP_001121696.1 4, 5 1343
          NR_033361.1 2, 3 1488
          XM_006713542.3 XP_006713605.1 5, 6 2055
          XM_006713543.3 XP_006713606.1 5, 6 2052
          XM_006713544.1 XP_006713607.1 5, 6 2180
          XM_006713550.3 XP_006713613.1 5, 6 1613
          XM_006713552.3 XP_006713615.1 2, 3 1235
          XM_006713553.3 XP_006713616.1 2, 3 2527
          XM_011512572.2 XP_011510874.1 6, 7 2215
          XM_011512573.2 XP_011510875.1 5, 6 2794
          XM_011512576.2 XP_011510878.1 5, 6 2048
          XM_011512579.2 XP_011510881.1 4, 5 1803
          XM_011512580.2 XP_011510882.1 7, 8 1940
          XM_011512581.2 XP_011510883.1 6, 7 1777
          XM_011512582.1 XP_011510884.1 2, 3 1569
          XM_011512583.2 XP_011510885.1 4, 5 1346
          XM_011512584.2 XP_011510886.1 6, 7 1665
          XM_017005931.1 XP_016861420.1 5, 6 1963
          XM_017005932.1 XP_016861421.1 6, 7 2202
          XM_017005933.1 XP_016861422.1 4, 5 1999
          XM_017005934.1 XP_016861423.1 8, 9 2013
          XM_017005935.1 XP_016861424.1 9, 10 2102
          XM_017005936.1 XP_016861425.1 6, 7 1776
          XM_017005937.1 XP_016861426.1 6, 7 1710
          XM_017005938.1 XP_016861427.1 8, 9 2076
          XM_017005939.1 XP_016861428.1 7, 8 1970
          XM_017005940.1 XP_016861429.1 4, 5 1348
          XM_017005941.1 XP_016861430.1 4, 5 1474
          GenBank mRNA AB018322.1 - 1, 2 509
          AK124671.1 - 2, 3 1489
          AK291448.1 - 4, 5 1761
          AK293408.1 - 2, 3 1025
          BC012480.1 - 1, 2 72
          BC030619.2 - 2, 3 1406
          BC034284.1 - 1, 2 92
          BC039859.1 - 4, 5 1849 5527623
          BC047517.1 - 2, 3 1406
          BX640712.1 - 2, 3 996
          CR749206.1 - 2, 3 1468

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0