Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYT1L SIRNA Assays ›
          Gene Symbol:
          MYT1L
          Gene Name:
          myelin transcription factor 1 like
          Gene Aliases:
          MRD39, NZF1, ZC2H2C2, ZC2HC4B
          Chromosome Location:
          Chr.2: 1789113 - 2331388 on Build GRCh38
          UniGene ID:
          Hs.434418
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22897

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYT1L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303052.1 NP_001289981.1 23 4121
          NM_015025.3 NP_055840.2 23 4115
          XM_011510318.1 XP_011508620.1 24 4246
          XM_011510319.1 XP_011508621.1 24 4356
          XM_011510320.2 XP_011508622.1 24 4287
          XM_011510321.2 XP_011508623.1 24 4237
          XM_011510322.2 XP_011508624.1 24 4050
          XM_011510323.1 XP_011508625.1 23 3969
          XM_011510324.1 XP_011508626.1 24 4246
          XM_011510325.1 XP_011508627.1 24 4240
          XM_011510326.1 XP_011508628.1 23 4183
          XM_011510327.1 XP_011508629.1 24 4246
          XM_011510328.2 XP_011508630.1 24 4246
          XM_017003604.1 XP_016859093.1 25 4335
          XM_017003605.1 XP_016859094.1 24 3914
          XM_017003606.1 XP_016859095.1 24 4350
          XM_017003607.1 XP_016859096.1 24 4281
          XM_017003609.1 XP_016859098.1 23 4168
          XM_017003610.1 XP_016859099.1 24 4267
          XM_017003611.1 XP_016859100.1 23 4177
          XM_017003612.1 XP_016859101.1 23 4287
          XM_017003613.1 XP_016859102.1 23 4287
          XM_017003614.1 XP_016859103.1 23 4126
          XM_017003615.1 XP_016859104.1 23 4183
          XM_017003616.1 XP_016859105.1 23 4183
          XM_017003617.1 XP_016859106.1 23 4177
          XM_017003618.1 XP_016859107.1 23 4177
          XM_017003619.1 XP_016859108.1 23 4287
          XM_017003620.1 XP_016859109.1 17 3236
          GenBank mRNA AB029029.2 - 24 3900
          AF036943.1 - 23 4016
          BC071612.1 - 5 598
          BC137272.1 - 23 3918 9021269
          BC137273.1 - 23 3918 9021270
          BC150281.1 - 24 3900 9007267
          BM715930.1 - 3 415

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          nervous system development
          cell differentiation
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0