Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015325.2 | NP_056140.1 | 13 | 5053 | ||
XM_011513999.2 | XP_011512301.1 | 13 | 5053 | |||
XM_017009285.1 | XP_016864774.1 | 14 | 5142 | |||
GenBank mRNA | AB023164.3 | - | 13 | 5032 | ||
AK024400.1 | - | 1 | 1983 | |||
AL832204.1 | - | 1 | 3524 | |||
BX538020.1 | - | 1 | 2542 | |||
CR749441.1 | - | 6 | 4324 |