Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFBR1 SIRNA Assays ›
          Gene Symbol:
          TGFBR1
          Gene Name:
          transforming growth factor beta receptor 1
          Gene Aliases:
          AAT5, ACVRLK4, ALK-5, ALK5, ESS1, LDS1, LDS1A, LDS2A, MSSE, SKR4, TGFR-1, tbetaR-I
          Chromosome Location:
          Chr.9: 99104038 - 99154192 on Build GRCh38
          UniGene ID:
          Hs.494622
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s229439

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TGFBR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130916.2 NP_001124388.1 4 781
          NM_001306210.1 NP_001293139.1 5 1024
          NM_004612.3 NP_004603.1 5 1012
          XM_011518948.2 XP_011517250.1 5 939
          XM_011518949.2 XP_011517251.1 5 867
          XM_011518950.2 XP_011517252.1 4 3969
          XM_017015063.1 XP_016870552.1 5 902
          GenBank mRNA AJ619019.2 - 5 983
          AK302059.1 - 5 1012
          AK302234.1 - 5 885
          BC071181.1 - 4 759 30344890
          L11695.1 - 5 971

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          skeletal system development
          angiogenesis
          in utero embryonic development
          kidney development
          blastocyst development
          epithelial to mesenchymal transition
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lens development in camera-type eye
          ventricular trabecula myocardium morphogenesis
          ventricular compact myocardium morphogenesis
          proepicardium development
          regulation of transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          cell cycle arrest
          signal transduction
          transforming growth factor beta receptor signaling pathway
          heart development
          positive regulation of cell proliferation
          germ cell migration
          male gonad development
          post-embryonic development
          anterior/posterior pattern specification
          regulation of epithelial to mesenchymal transition
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          signal transduction by protein phosphorylation
          collagen fibril organization
          positive regulation of cell growth
          positive regulation of cell migration
          negative regulation of transforming growth factor beta receptor signaling pathway
          regulation of protein ubiquitination
          negative regulation of chondrocyte differentiation
          activin receptor signaling pathway
          intracellular signal transduction
          wound healing
          endothelial cell activation
          extracellular structure organization
          regulation of protein binding
          endothelial cell migration
          positive regulation of transcription, DNA-templated
          thymus development
          neuron fate commitment
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          mesenchymal cell differentiation
          artery morphogenesis
          cell motility
          positive regulation of cellular component movement
          positive regulation of filopodium assembly
          positive regulation of stress fiber assembly
          positive regulation of protein kinase B signaling
          parathyroid gland development
          palate development
          pharyngeal system development
          regulation of cardiac muscle cell proliferation
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          positive regulation of SMAD protein import into nucleus
          ventricular septum morphogenesis
          coronary artery morphogenesis
          response to cholesterol
          cellular response to transforming growth factor beta stimulus
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of occluding junction disassembly
          positive regulation of apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type I
          receptor signaling protein activity
          type II transforming growth factor beta receptor binding
          protein binding
          ATP binding
          growth factor binding
          SMAD binding
          metal ion binding
          transforming growth factor beta binding
          I-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0