Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SETX SIRNA Assays ›
          Gene Symbol:
          SETX
          Gene Name:
          senataxin
          Gene Aliases:
          ALS4, AOA2, SCAR1, bA479K20.2
          Chromosome Location:
          Chr.9: 132261440 - 132356726 on Build GRCh38
          UniGene ID:
          Hs.460317
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22951

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SETX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015046.5 NP_055861.3 8, 9 1201
          XM_005272171.1 XP_005272228.1 8, 9 1202
          XM_005272172.2 XP_005272229.1 9, 10 1300
          XM_005272173.2 XP_005272230.1 8, 9 1338
          XM_011518404.2 XP_011516706.1 9, 10 1342
          XM_011518405.2 XP_011516707.1 8, 9 1360
          XM_011518406.2 XP_011516708.1 8, 9 1202
          XM_011518407.1 XP_011516709.1 8, 9 1202
          XM_011518408.2 XP_011516710.1 8, 9 1202
          XM_017014495.1 XP_016869984.1 8, 9 1273
          GenBank mRNA AB014525.2 - 6, 7 978
          AK125448.1 - 8, 9 1338
          AK307608.1 - 8, 9 1185
          AY362728.1 - 6, 7 1025
          BC137350.1 - 7, 8 1135 9021348
          BX537849.1 - 8, 9 1185
          BX538166.1 - 8, 9 1204

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_198033, AK044730, AK048354.

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          double-strand break repair
          DNA recombination
          DNA-templated transcription, termination
          termination of RNA polymerase II transcription
          mRNA splice site selection
          RNA processing
          cellular response to DNA damage stimulus
          spermatogenesis
          circadian rhythm
          fibroblast growth factor receptor signaling pathway
          positive regulation of neuron projection development
          DNA duplex unwinding
          positive regulation of RNA splicing
          cellular response to oxidative stress
          negative regulation of apoptotic process
          protein kinase B signaling
          cellular response to fibroblast growth factor stimulus
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of DNA-templated transcription, termination
          cellular response to hydrogen peroxide
          cellular response to retinoic acid
          positive regulation of DNA-templated transcription, initiation
          positive regulation of termination of RNA polymerase II transcription, poly(A)-coupled
          transcription termination site sequence-specific DNA binding
          DNA binding
          DNA helicase activity
          protein binding
          ATP binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0