Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013382.5 | NP_037514.2 | 11 | 1414 | ||
XM_011536675.2 | XP_011534977.1 | 11 | 1414 | |||
XM_011536676.2 | XP_011534978.1 | 10 | 1327 | |||
XM_011536677.2 | XP_011534979.1 | 7 | 956 | |||
XM_011536679.2 | XP_011534981.1 | 7 | 616 | |||
XM_017021244.1 | XP_016876733.1 | 10 | 1248 | |||
XM_017021245.1 | XP_016876734.1 | 7 | 702 | |||
GenBank mRNA | AB209114.1 | - | 11 | 1236 | ||
AF105020.1 | - | 11 | 1417 | |||
AK023804.1 | - | 5 | 2005 | |||
AK025962.1 | - | 1 | 33 | |||
AY090480.1 | - | 11 | 1417 | |||
BC031651.1 | - | 11 | 1304 | 5169145 | ||
BM127609.1 | - | 5 | 338 | |||
CR627447.1 | - | 11 | 984 |