Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TBC1D3I SIRNA Assays ›
          Gene Symbol:
          TBC1D3I LOC101060389 TBC1D3 TBC1D3B TBC1D3C TBC1D3E TBC1D3F TBC1D3G TBC1D3H TBC1D3K TBC1D3L
          Gene Name:
          TBC1 domain family member 3I
          TBC1 domain family member-like
          TBC1 domain family member 3
          TBC1 domain family member 3B
          TBC1 domain family member 3C
          TBC1 domain family member 3E
          TBC1 domain family member 3F
          TBC1 domain family member 3G
          TBC1 domain family member 3H
          TBC1 domain family member 3K
          TBC1 domain family member 3L
          Gene Aliases:
          -
          TBC1D3D
          PRC17, TBC1D3A, TBC1D3F
          PRC17, TBC1D3I, TBC1D3L
          TBC1D3D
          TBC1DE3
          TBC1D3
          -
          -
          -
          -
          Chromosome Location:
          Chr.17: 36253639 - 36264601 on Build GRCh38
          Chromosome Location:
          Chr.17: 38004033 - 38014908 on Build GRCh38
          Chromosome Location:
          Chr.17: 38181476 - 38192555 on Build GRCh38
          Chromosome Location:
          Chr.17: 36165681 - 36176624 on Build GRCh38
          Chromosome Location:
          Chr.17: 38057693 - 38068634 on Build GRCh38
          Chromosome Location:
          Chr.17: 38119293 - 38138868 on Build GRCh38
          Chromosome Location:
          Chr.17: 36428612 - 36439523 on Build GRCh38
          Chromosome Location:
          Chr.17: 36322242 - 36334759 on Build GRCh38
          Chromosome Location:
          Chr.17: 36377475 - 36388448 on Build GRCh38
          Chromosome Location:
          Chr.17: 37923805 - 37935371 on Build GRCh38
          Chromosome Location:
          Chr.17: 37978155 - 37990691 on Build GRCh38
          UniGene ID:
          Hs.454716
          Hs.454716
          Hs.454716
          Hs.105891
          Hs.735975
          Hs.454716
          Hs.617010
          Hs.454716
          Hs.105891
          Hs.454716
          Hs.454716
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s229691

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TBC1D3I
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291463.1 NP_001278392.1 6 393
          XM_006722223.3 XP_006722286.1 6 664
          XM_006722224.3 XP_006722287.1 5 1909
          XM_006722226.3 XP_006722289.1 5 1909
          XM_011524183.2 XP_011522485.1 5 2092
          XM_011524184.2 XP_011522486.1 5 2092
          XM_011524185.2 XP_011522487.1 5 2092
          GenBank mRNA BC078140.1 - 6 394 5742850
          Gene Symbol: LOC101060389
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291465.1 NP_001278394.1 6 393
          XM_006722248.2 XP_006722311.1 5 1899
          XM_006722249.2 XP_006722312.1 5 1899
          XM_006722250.2 XP_006722313.1 5 1899
          XM_006722252.3 XP_006722315.1 5 1716
          XM_006722253.2 XP_006722316.1 5 1899
          XM_011524176.2 XP_011522478.1 6 506
          XM_017024010.1 XP_016879499.1 5 1899
          GenBank mRNA BI088323.1 - 4 156
          Gene Symbol: TBC1D3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123391.3 NP_001116863.3 6 405
          GenBank mRNA AB449884.1 - 5 293
          AK302064.1 - 7 631
          AK303259.1 - 5 425
          AX775941.1 - 8 654
          AX775943.1 - 8 654
          Gene Symbol: TBC1D3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001417.6 NP_001001417.6 6 439
          XM_005257980.4 XP_005258037.2 6 569
          GenBank mRNA AF540953.1 - 5 293
          AK292257.1 - 6 410
          Gene Symbol: TBC1D3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001418.6 NP_001001418.5 6 433
          XM_006721904.2 XP_006721967.1 5 1899
          XM_006721905.2 XP_006721968.1 5 1899
          XM_006721907.3 XP_006721970.1 5 1716
          XM_006721908.2 XP_006721971.1 5 1899
          XM_011524814.2 XP_011523116.1 6 506
          XM_017024646.1 XP_016880135.1 5 1899
          GenBank mRNA AK313329.1 - 6 433
          Gene Symbol: TBC1D3E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AL136860.1 - 6 391
          BC047739.1 - 7 604
          Gene Symbol: TBC1D3F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032258.4 NP_115634.3 6 407
          GenBank mRNA AK225985.1 - 6 398
          AK292344.1 - 6 450
          BC075809.1 - 6 439 5170064
          Gene Symbol: TBC1D3G
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291462.1 NP_001278391.1 6 393
          XM_005276914.3 XP_005276971.1 6 2218
          GenBank mRNA AK292308.1 - 6 443
          AK301823.1 - 6 392
          BC071680.1 - 6 393 5165385
          Gene Symbol: TBC1D3H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123392.3 NP_001116864.3 6 407
          XM_006722055.1 XP_006722118.1 5 1047
          XM_006722056.1 XP_006722119.1 5 1047
          XM_011525172.1 XP_011523474.1 5 1230
          XM_011525173.2 XP_011523475.1 6 601
          XM_011525174.1 XP_011523476.1 5 1230
          XM_011525175.1 XP_011523477.1 5 1230
          XM_011525176.1 XP_011523478.1 5 1230
          GenBank mRNA CB131287.1 - 5 243
          DC398423.1 - 6 408
          Gene Symbol: TBC1D3K
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291464.1 NP_001278393.1 6 393
          XM_006722235.2 XP_006722298.1 5 2671
          XM_006722236.1 XP_006722299.1 6 1243
          XM_006722237.2 XP_006722300.1 5 2488
          XM_006722239.2 XP_006722302.1 5 2488
          XM_011524172.1 XP_011522474.1 5 2671
          XM_011524173.1 XP_011522475.1 5 2671
          XM_011524174.1 XP_011522476.1 5 2671
          GenBank mRNA DC397859.1 - 6 426
          Gene Symbol: TBC1D3L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291459.1 NP_001278388.1 6 407
          XM_006722242.3 XP_006722305.1 5 3647
          XM_006722243.2 XP_006722306.1 5 1899
          XM_006722244.2 XP_006722307.1 5 1899
          XM_006722246.3 XP_006722309.1 5 1047
          XM_006722247.2 XP_006722310.1 5 1899
          XM_011524175.2 XP_011522477.1 6 506
          GenBank mRNA AK095385.1 - 5 1047
          AK122833.1 - 2 1718
          AX775949.1 - 5 1047
          AX775951.1 - 5 1047

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          regulation of cilium assembly
          intracellular protein transport
          regulation of vesicle fusion
          activation of GTPase activity
          GTPase activator activity
          Rab GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0