Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s229971
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
7
1608
NM_001174093.1
NP_001167564.1
6
1268
NM_001174094.1
NP_001167565.1
7
1605
NM_001174095.1
NP_001167566.1
6
1127
NM_001174096.1
NP_001167567.1
7
1331
NM_001323638.1
NP_001310567.1
11
1978
NM_001323641.1
NP_001310570.1
7
1558
NM_001323642.1
NP_001310571.1
9
1860
NM_001323643.1
NP_001310572.1
9
1638
NM_001323644.1
NP_001310573.1
6
1357
NM_001323645.1
NP_001310574.1
8
1701
NM_001323646.1
NP_001310575.1
10
1939
NM_001323647.1
NP_001310576.1
8
1471
NM_001323648.1
NP_001310577.1
7
1411
NM_001323649.1
NP_001310578.1
8
1474
NM_001323650.1
NP_001310579.1
10
1706
NM_001323651.1
NP_001310580.1
8
1374
NM_001323652.1
NP_001310581.1
9
1523
NM_001323653.1
NP_001310582.1
9
1630
NM_001323654.1
NP_001310583.1
9
1517
NM_001323655.1
NP_001310584.1
9
1627
NM_001323656.1
NP_001310585.1
7
1302
NM_001323657.1
NP_001310586.1
7
1305
NM_001323658.1
NP_001310587.1
7
1144
NM_001323659.1
NP_001310588.1
7
1362
NM_001323660.1
NP_001310589.1
7
1359
NM_001323661.1
NP_001310590.1
8
1505
NM_001323662.1
NP_001310591.1
9
1907
NM_001323663.1
NP_001310592.1
10
1846
NM_001323664.1
NP_001310593.1
9
1830
NM_001323665.1
NP_001310594.1
8
1748
NM_001323666.1
NP_001310595.1
7
1688
NM_001323671.1
NP_001310600.1
8
1584
NM_001323672.1
NP_001310601.1
9
1636
NM_001323673.1
NP_001310602.1
8
1581
NM_001323674.1
NP_001310603.1
5
1106
NM_001323675.1
NP_001310604.1
5
1216
NM_001323676.1
NP_001310605.1
7
1441
NM_001323677.1
NP_001310606.1
7
1438
NM_001323678.1
NP_001310607.1
5
1383
NM_030751.5
NP_110378.3
7
1328
XM_006717498.2
XP_006717561.1
10
1766
XM_011519643.2
XP_011517945.1
10
2069
XM_017016597.1
XP_016872086.1
10
1668
XM_017016598.1
XP_016872087.1
10
4798
XM_017016599.1
XP_016872088.1
9
4636
XM_017016600.1
XP_016872089.1
11
4877
XM_017016601.1
XP_016872090.1
10
4795
XM_017016602.1
XP_016872091.1
11
4874
XM_017016603.1
XP_016872092.1
10
4712
GenBank mRNA
AK091478.1
-
6
1228
AK294775.1
-
7
1523
AK296244.1
-
7
1251
AK299541.1
-
4
865
AK299666.1
-
7
1362
AK300830.1
-
6
1075
AK307510.1
-
8
1334
AK314683.1
-
7
1289
AL831979.1
-
9
1586
BC009572.1
-
7
1284
BC065705.1
-
7
1289
BC107781.1
-
7
1284
BC112392.1
-
7
1284
40036600
BX647794.1
-
11
1979
M81699.1
-
1
200
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0