Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-DQA1 SIRNA Assays ›
          Gene Symbol:
          HLA-DQA1
          Gene Name:
          major histocompatibility complex, class II, DQ alpha 1
          Gene Aliases:
          CD, CELIAC1, DQ-A1, GSE, HLA-DQA
          Chromosome Location:
          Chr.6: 32637403 - 32654846 on Build GRCh38
          UniGene ID:
          Hs.387679
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s230157

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-DQA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002122.3 NP_002113.2 2, 3 384
          XM_006715079.3 XP_006715142.1 2, 3 387
          GenBank mRNA AB209628.1 - 2, 3 359
          AF012768.1 - 1, 2 228
          AK130598.1 - 2, 3 379
          AK130811.1 - 2, 3 384
          AK130838.1 - 2, 3 381
          AK313975.1 - 2, 3 386
          BC008585.1 - 2, 3 362 4339446
          BC051832.1 - 3, 4 1357
          BC073977.1 - 2, 3 365 6014803
          BC125044.1 - 2, 3 378 40116009
          BC125045.1 - 2, 3 378 40116010
          BC157865.1 - 2, 3 350 9057084
          BC171877.1 - 2, 3 335 9054531
          CR450297.1 - 2, 3 328
          DQ284436.1 - 2, 3 393
          DQ284437.1 - 2, 3 392
          DQ284438.1 - 2, 3 393
          DQ284439.1 - 2, 3 392
          DQ284440.1 - 2, 3 389
          J00199.1 - 1, 2 215
          JF895477.1 - 2, 3 331
          L34082.1 - 1, 2 262
          L34083.1 - 1, 2 262
          L34084.1 - 1, 2 262
          L34085.1 - 1, 2 262
          L34086.1 - 1, 2 262
          L34087.1 - 1, 2 259
          L34088.1 - 1, 2 262
          L34090.1 - 1, 2 259
          L34091.1 - 1, 2 259
          L34092.1 - 1, 2 259
          L34093.1 - 1, 2 259
          L34094.1 - 1, 2 259
          L42625.1 - 1, 2 262
          M11124.1 - 1, 2 262
          M16995.1 - 2, 3 328
          M20431.1 - 2, 3 265
          M20506.1 - 1, 2 227
          M26041.1 - 2, 3 371
          M33906.1 - 2, 3 371
          M34996.1 - 2, 3 295
          U77589.1 - 2, 3 331
          X00033.1 - 2, 3 347
          X00370.1 - 2, 3 338
          X00452.1 - 2, 3 353

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          T cell costimulation
          T cell receptor signaling pathway
          interferon-gamma-mediated signaling pathway
          MHC class II receptor activity
          peptide antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0