Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC8A1 SIRNA Assays ›
          Gene Symbol:
          SLC8A1
          Gene Name:
          solute carrier family 8 member A1
          Gene Aliases:
          NCX1
          Chromosome Location:
          Chr.2: 40112146 - 40512452 on Build GRCh38
          UniGene ID:
          Hs.31961
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s230227

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC8A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001112800.1 NP_001106271.1 6, 7 2183
          NM_001112801.1 NP_001106272.1 5, 6 2159
          NM_001112802.1 NP_001106273.1 5, 6 2177
          NM_001252624.1 NP_001239553.1 6, 7 2174
          NM_021097.2 NP_066920.1 7, 8 2198
          XM_005264514.3 XP_005264571.1 5, 6 2272
          XM_006712081.2 XP_006712144.1 8, 9 2380
          XM_006712082.3 XP_006712145.1 9, 10 2482
          XM_006712083.3 XP_006712146.1 8, 9 2299
          XM_006712084.3 XP_006712147.1 9, 10 2580
          XM_006712085.3 XP_006712148.1 8, 9 3202
          XM_011533050.1 XP_011531352.1 6, 7 2174
          XM_011533054.2 XP_011531356.1 8, 9 2216
          XM_011533055.2 XP_011531357.1 8, 9 2343
          XM_011533056.1 XP_011531358.1 8, 9 2216
          XM_011533057.2 XP_011531359.2 5, 6 2259
          XM_017004745.1 XP_016860234.1 8, 9 2364
          XM_017004746.1 XP_016860235.1 8, 9 2217
          XM_017004747.1 XP_016860236.1 6, 7 2383
          XM_017004748.1 XP_016860237.1 8, 9 2285
          XM_017004749.1 XP_016860238.1 9, 10 2485
          XM_017004750.1 XP_016860239.1 10, 11 2719
          XM_017004751.1 XP_016860240.1 10, 11 2716
          XM_017004752.1 XP_016860241.1 9, 10 2520
          XM_017004753.1 XP_016860242.1 8, 9 2216
          XM_017004754.1 XP_016860243.1 7, 8 2320
          XM_017004755.1 XP_016860244.1 6, 7 2307
          XM_017004756.1 XP_016860245.1 6, 7 2387
          XM_017004757.1 XP_016860246.1 6, 7 2382
          XM_017004758.1 XP_016860247.1 6, 7 2361
          XM_017004759.1 XP_016860248.1 6, 7 2322
          XM_017004760.1 XP_016860249.1 6, 7 2296
          XM_017004761.1 XP_016860250.1 6, 7 2339
          XM_017004762.1 XP_016860251.1 5, 6 2293
          XM_017004763.1 XP_016860252.1 5, 6 2275
          GenBank mRNA AB209075.1 - 6, 7 488
          AF108388.1 - 5, 6 2135
          AF108389.1 - 4, 5 2066
          AF128524.1 - 5, 6 2157
          AK291696.1 - 5, 6 2177
          BC098285.1 - 7, 8 408
          BC098308.1 - 6, 7 392
          BX648299.1 - 5, 6 2257
          M91368.1 - 2254
          M96368.1 - 7, 8 2215

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          regulation of heart rate
          regulation of sodium ion transport
          ion transport
          cellular sodium ion homeostasis
          muscle contraction
          response to nutrient
          response to glucose
          regulation of cell communication by electrical coupling
          positive regulation of fibroblast migration
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of cardiac muscle contraction by calcium ion signaling
          vascular smooth muscle contraction
          telencephalon development
          positive regulation of bone mineralization
          response to ATP
          cellular response to reactive oxygen species
          sodium ion transmembrane transport
          response to immobilization stress
          response to muscle stretch
          response to drug
          response to hydrogen peroxide
          relaxation of smooth muscle
          negative regulation of cytosolic calcium ion concentration
          cardiac muscle cell development
          calcium ion homeostasis
          relaxation of cardiac muscle
          cardiac muscle contraction
          cytosolic calcium ion transport
          calcium ion transport into cytosol
          calcium ion import
          calcium ion transmembrane transport
          cellular response to caffeine
          cellular response to cAMP
          sodium ion export
          cellular response to hypoxia
          membrane depolarization during cardiac muscle cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          sodium ion import
          positive regulation of the force of heart contraction
          calcium ion export
          regulation of cardiac conduction
          calcium:sodium antiporter activity
          calcium ion binding
          protein binding
          calmodulin binding
          cytoskeletal protein binding
          ankyrin binding
          ion channel binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-26z5w:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0