Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000723.4 | NP_000714.3 | 3 | 481 | ||
NM_199247.2 | NP_954855.1 | 3 | 481 | |||
NM_199248.2 | NP_954856.1 | 3 | 481 | |||
XM_005257645.2 | XP_005257702.1 | 3 | 416 | |||
XM_005257646.2 | XP_005257703.1 | 3 | 416 | |||
XM_006722072.2 | XP_006722135.1 | 2 | 147 | |||
XM_017025024.1 | XP_016880513.1 | 3 | 286 | |||
XM_017025025.1 | XP_016880514.1 | 2 | 147 | |||
XM_017025026.1 | XP_016880515.1 | 2 | 177 | |||
XM_017025027.1 | XP_016880516.1 | 3 | 394 | |||
XM_017025028.1 | XP_016880517.1 | 3 | 840 | |||
XM_017025029.1 | XP_016880518.1 | 2 | 177 | |||
XM_017025030.1 | XP_016880519.1 | 3 | 416 | |||
GenBank mRNA | AB054985.1 | - | 3 | 434 | ||
AB209176.1 | - | 1 | 2392 | |||
AK127283.1 | - | 1 | 1160 | |||
AK289729.1 | - | 3 | 447 | |||
AK300666.1 | - | 2 | 151 | |||
AK312385.1 | - | 3 | 480 | |||
AY393857.1 | - | 3 | 273 | |||
BC037311.1 | - | 3 | 418 | 5262182 | ||
BP217857.1 | - | 3 | 481 | |||
L06111.1 | - | 3 | 270 | |||
L06112.1 | - | 3 | 270 | |||
M76560.1 | - | 3 | 273 | |||
M92301.1 | - | 3 | 422 | |||
M92302.1 | - | 3 | 422 | |||
M92303.1 | - | 3 | 422 |