Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199287.2 | NP_954981.1 | 2 | 280 | ||
XM_011524738.2 | XP_011523040.1 | 2 | 187 | |||
XM_017024573.1 | XP_016880062.1 | 2 | 314 | |||
GenBank mRNA | BC009369.2 | - | 2 | 264 | 4110481 | |
BM548065.1 | - | 2 | 319 | |||
FJ649645.1 | - | 2 | 1970 |