siRNA Details
Gene Symbol: BZW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207067.1 | NP_001193996.1 | 9 | 1344 | ||
NM_001207069.1 | NP_001193998.1 | 9 | 1024 | |||
NM_001321688.1 | NP_001308617.1 | 9 | 924 | |||
NM_001321690.1 | NP_001308619.1 | 9 | 1038 | |||
NM_001321691.1 | NP_001308620.1 | 9 | 968 | |||
NM_001321693.1 | NP_001308622.1 | 8 | 791 | |||
NM_001321694.1 | NP_001308623.1 | 9 | 899 | |||
XM_017005393.1 | XP_016860882.1 | 6 | 637 | |||
GenBank mRNA | AK096489.1 | - | 8 | 901 | ||
AK223245.1 | - | 9 | 953 | |||
AK297227.1 | - | 9 | 1037 | |||
AK301511.1 | - | 9 | 1024 | |||
AL833518.1 | - | 9 | 1300 | |||
BC026303.1 | - | 9 | 953 | 4284614 | ||
D13630.1 | - | 9 | 969 |

More Information
Panther Classification:
Molecular Function -





