Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124767.1 | NP_001118239.1 | 1 | 256 | ||
XM_011533729.2 | XP_011532031.1 | 1 | 372 | |||
GenBank mRNA | AK095910.1 | - | 1 | 256 | ||
BC017996.1 | - | 1 | 256 | |||
BG031013.1 | - | 1 | 246 |