Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052999.3 | NP_443725.3 | 4 | 950 | ||
NM_181268.2 | NP_851785.2 | 4 | 934 | |||
NM_181269.2 | NP_851786.1 | 4 | 655 | |||
NM_181270.2 | NP_851787.1 | 4 | 639 | |||
NM_181271.2 | NP_851788.1 | 3 | 514 | |||
NM_181272.2 | NP_851789.1 | 3 | 496 | |||
NM_181283.2 | NP_851800.1 | 3 | 480 | |||
NM_181296.2 | NP_851813.1 | 2 | 381 | |||
GenBank mRNA | AF278576.1 | - | 4 | 554 | ||
AF278577.1 | - | 4 | 570 | |||
AY174118.1 | - | 4 | 659 | |||
AY174119.1 | - | 4 | 643 | |||
AY174120.1 | - | 3 | 518 | |||
AY174121.1 | - | 3 | 500 | |||
AY174122.1 | - | 3 | 484 | |||
AY174123.1 | - | 3 | 655 | |||
AY174124.1 | - | 3 | 639 | |||
AY174125.1 | - | 5 | 793 | |||
AY174126.1 | - | 5 | 777 | |||
AY174127.1 | - | 2 | 347 | |||
AY174128.1 | - | 4 | 623 | |||
AY174129.1 | - | 5 | 646 | |||
AY174130.1 | - | 5 | 818 | |||
AY174131.1 | - | 3 | 475 | |||
AY174132.1 | - | 3 | 459 | |||
AY174133.1 | - | 2 | 385 | |||
AY174134.1 | - | 4 | 705 | |||
AY174135.1 | - | 5 | 883 | |||
AY174136.1 | - | 5 | 867 | |||
AY174137.1 | - | 4 | 685 | |||
AY174138.1 | - | 4 | 669 | |||
DQ309766.1 | - | 5 | 768 | |||
DQ355985.1 | - | 3 | 506 |
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202509.1 | NP_001189438.1 | 3 | 523 | ||
NM_001204098.1 | NP_001191027.1 | 4 | 603 | |||
NM_001204099.1 | NP_001191028.1 | 4 | 682 |