Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112734.3 | NP_001106205.1 | 4 | 1115 | ||
NM_001166012.2 | NP_001159484.1 | 5 | 1319 | |||
NM_001288711.1 | NP_001275640.1 | 2 | 412 | |||
XM_005258183.4 | XP_005258240.1 | 4 | 1084 | |||
XM_006722371.3 | XP_006722434.1 | 5 | 1248 | |||
XM_011525789.2 | XP_011524091.1 | 1 | 313 | |||
XM_017025515.1 | XP_016881004.1 | 5 | 1267 | |||
XM_017025516.1 | XP_016881005.1 | 4 | 1100 | |||
XM_017025517.1 | XP_016881006.1 | 4 | 1094 | |||
XM_017025518.1 | XP_016881007.1 | 4 | 1166 | |||
XM_017025519.1 | XP_016881008.1 | 4 | 1071 | |||
XM_017025520.1 | XP_016881009.1 | 3 | 965 | |||
XM_017025521.1 | XP_016881010.1 | 3 | 918 | |||
XM_017025522.1 | XP_016881011.1 | 3 | 897 | |||
GenBank mRNA | AF303374.1 | - | 1 | 537 | ||
AK303447.1 | - | 5 | 1303 | |||
AY234408.1 | - | 4 | 1078 | |||
DA653861.1 | - | 2 | 397 |