Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP20-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128077.1 | NP_001121549.1 | 1 | 126 | ||
GenBank mRNA | AB096958.1 | - | 1 | 126 |