Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015086.1 | NP_055901.2 | 2 | 1146 | ||
XM_011538055.2 | XP_011536357.1 | 3 | 1751 | |||
GenBank mRNA | AB018292.1 | - | 2 | 1030 | ||
BC136398.1 | - | 2 | 1056 | 9020385 | ||
BC136399.1 | - | 2 | 1056 | 9020386 |