Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TBC1D3E SIRNA Assays ›
          Gene Symbol:
          TBC1D3E LOC101060389 TBC1D3 TBC1D3B TBC1D3C TBC1D3F TBC1D3G TBC1D3H TBC1D3I TBC1D3J TBC1D3K TBC1D3L TBC1D3P2
          Gene Name:
          TBC1 domain family member 3E
          TBC1 domain family member-like
          TBC1 domain family member 3
          TBC1 domain family member 3B
          TBC1 domain family member 3C
          TBC1 domain family member 3F
          TBC1 domain family member 3G
          TBC1 domain family member 3H
          TBC1 domain family member 3I
          TBC1 domain family member 3J
          TBC1 domain family member 3K
          TBC1 domain family member 3L
          TBC1 domain family member 3 pseudogene 2
          Gene Aliases:
          TBC1DE3
          TBC1D3D
          PRC17, TBC1D3A, TBC1D3F
          PRC17, TBC1D3I, TBC1D3L
          TBC1D3D
          TBC1D3
          -
          -
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.17: 38119293 - 38138868 on Build GRCh38
          Chromosome Location:
          Chr.17: 38004033 - 38014908 on Build GRCh38
          Chromosome Location:
          Chr.17: 38181476 - 38192555 on Build GRCh38
          Chromosome Location:
          Chr.17: 36165681 - 36176624 on Build GRCh38
          Chromosome Location:
          Chr.17: 38057693 - 38068634 on Build GRCh38
          Chromosome Location:
          Chr.17: 36428612 - 36439523 on Build GRCh38
          Chromosome Location:
          Chr.17: 36322242 - 36334759 on Build GRCh38
          Chromosome Location:
          Chr.17: 36377475 - 36388448 on Build GRCh38
          Chromosome Location:
          Chr.17: 36253639 - 36264601 on Build GRCh38
          Chromosome Location:
          Chr.17: 36450649 - 36458000 on Build GRCh38
          Chromosome Location:
          Chr.17: 37923805 - 37935371 on Build GRCh38
          Chromosome Location:
          Chr.17: 37978155 - 37990691 on Build GRCh38
          Chromosome Location:
          Chr.17: 62264705 - 62275655 on Build GRCh38
          UniGene ID:
          Hs.454716
          Hs.454716
          Hs.454716
          Hs.105891
          Hs.735975
          Hs.617010
          Hs.454716
          Hs.105891
          Hs.454716
          -
          Hs.454716
          Hs.454716
          Hs.448884
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s230582

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TBC1D3E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291466.1 NP_001278395.1 11, 12 918
          XM_006722254.1 XP_006722317.1 14, 15 1267
          GenBank mRNA AL136860.1 - 11, 12 918
          BC047739.1 - 12, 13 1131
          Gene Symbol: LOC101060389
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291465.1 NP_001278394.1 11, 12 920
          XM_006722248.2 XP_006722311.1 10, 11 2426
          XM_006722250.2 XP_006722313.1 10, 11 2426
          XM_006722253.2 XP_006722316.1 10, 11 2426
          XM_011524176.2 XP_011522478.1 11, 12 1033
          GenBank mRNA BI088323.1 - 9, 10 683
          Gene Symbol: TBC1D3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123391.3 NP_001116863.3 11, 12 932
          GenBank mRNA AB449884.1 - 10, 11 820
          AK302064.1 - 12, 13 1158
          AK303259.1 - 10, 11 952
          AX775941.1 - 13, 14 1181
          AX775943.1 - 13, 14 1181
          Gene Symbol: TBC1D3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001417.6 NP_001001417.6 11, 12 966
          XM_005257980.4 XP_005258037.2 11, 12 1096
          GenBank mRNA AF540953.1 - 10, 11 820
          AK292257.1 - 11, 12 937
          Gene Symbol: TBC1D3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001418.6 NP_001001418.5 11, 12 960
          XM_006721904.2 XP_006721967.1 10, 11 2426
          XM_006721908.2 XP_006721971.1 10, 11 2426
          XM_011524814.2 XP_011523116.1 11, 12 1033
          XM_017024646.1 XP_016880135.1 10, 11 2426
          Gene Symbol: TBC1D3F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032258.4 NP_115634.3 11, 12 934
          GenBank mRNA AK225985.1 - 11, 12 925
          Gene Symbol: TBC1D3G
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291462.1 NP_001278391.1 11, 12 920
          XM_005276914.3 XP_005276971.1 11, 12 2745
          GenBank mRNA AK292308.1 - 12, 13 970
          AK301823.1 - 9, 10 732
          BC071680.1 - 11, 12 920 5165385
          Gene Symbol: TBC1D3H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123392.3 NP_001116864.3 11, 12 934
          XM_006722055.1 XP_006722118.1 10, 11 1574
          XM_011525172.1 XP_011523474.1 10, 11 1757
          XM_011525173.2 XP_011523475.1 11, 12 1128
          XM_011525175.1 XP_011523477.1 10, 11 1757
          XM_011525176.1 XP_011523478.1 10, 11 1757
          GenBank mRNA BM844606.1 - 2, 3 132
          BX281608.1 - 5, 6 380
          Gene Symbol: TBC1D3I
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291463.1 NP_001278392.1 11, 12 920
          XM_006722223.3 XP_006722286.1 11, 12 1191
          XM_006722224.3 XP_006722287.1 10, 11 2436
          XM_011524184.2 XP_011522486.1 10, 11 2619
          XM_011524185.2 XP_011522487.1 10, 11 2619
          Gene Symbol: TBC1D3J
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017025494.1 XP_016880983.1 9, 10 736
          Gene Symbol: TBC1D3K
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291464.1 NP_001278393.1 11, 12 920
          XM_006722235.2 XP_006722298.1 10, 11 3198
          XM_006722236.1 XP_006722299.1 11, 12 1770
          XM_006722237.2 XP_006722300.1 10, 11 3015
          XM_011524173.1 XP_011522475.1 10, 11 3198
          XM_011524174.1 XP_011522476.1 10, 11 3198
          Gene Symbol: TBC1D3L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291459.1 NP_001278388.1 11, 12 934
          XM_006722242.3 XP_006722305.1 10, 11 4174
          XM_006722244.2 XP_006722307.1 10, 11 2426
          XM_006722247.2 XP_006722310.1 10, 11 2426
          XM_011524175.2 XP_011522477.1 11, 12 1033
          GenBank mRNA AK122833.1 - 7, 8 2245
          Gene Symbol: TBC1D3P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK301746.1 - 11, 12 880

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          regulation of cilium assembly
          intracellular protein transport
          regulation of vesicle fusion
          activation of GTPase activity
          GTPase activator activity
          Rab GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0